Skip to Content
Merck
All Photos(1)

Documents

46130

Supelco

4-Aminoazobenzene

analytical standard

Synonym(s):

4-Phenylazoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4NH2
CAS Number:
Molecular Weight:
197.24
Colour Index Number:
11000
Beilstein:
1913042
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

>360 °C (lit.)

mp

123-126 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(cc1)\N=N/c2ccccc2

InChI

1S/C12H11N3/c13-10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,13H2/b15-14+

InChI key

QPQKUYVSJWQSDY-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Gam T Le et al.
Visual neuroscience, 29(4-5), 219-228 (2012-09-28)
Vitamin A deficiency causes impaired vision and blindness in millions of children around the world. Previous studies in zebrafish have demonstrated that retinoic acid (RA), the acid form of vitamin A, plays a vital role in early eye development. The
Frederic Delbecq et al.
Journal of colloid and interface science, 384(1), 94-98 (2012-07-24)
A "light-triggerable" azobenzene amine derivative (additive 1) was synthesized and then introduced into organogels of 12-hydroxystearic acid (HSA) in the molar ratio of 1:3. The organogels (HSA/1) consisting of additive 1 and HSA were analyzed by (1)H nuclear magnetic resonance
Kristen Alexa et al.
PloS one, 4(12), e8261-e8261 (2009-12-17)
We have isolated and characterized a novel zebrafish pancreas mutant. Mutant embryos lack expression of isl1 and sst in the endocrine pancreas, but retain isl1 expression in the CNS. Non-endocrine endodermal gene expression is less affected in the mutant, with
Ana Neto et al.
Development (Cambridge, England), 139(2), 301-311 (2011-12-02)
Vertebrate odd-skipped related genes (Osr) have an essential function during the formation of the intermediate mesoderm (IM) and the kidney structures derived from it. Here, we show that these genes are also crucial for limb bud formation in the adjacent
'Passe-partout effect' in positive patch test reactions: a novel pattern of edge effect.
Esen Ozkaya
Contact dermatitis, 61(4), 245-247 (2009-10-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service