Skip to Content
Merck
All Photos(4)

Key Documents

242284

Sigma-Aldrich

Aniline

ACS reagent, ≥99.5%

Synonym(s):

Aminobenzene, Benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NH2
CAS Number:
Molecular Weight:
93.13
Beilstein:
605631
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030407
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
184 °C (lit.)
vapor pressure:
0.7 mmHg ( 25 °C)

grade

ACS reagent

Quality Level

vapor density

3.22 (185 °C, vs air)

vapor pressure

0.7 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

1139 °F

expl. lim.

11 %

dilution

(for analytical testing)

impurities

Hydrocarbons, passes test
Nitrobenzene, passes test (lim. ~0.001%)
≤0.01% chlorobenzene

ign. residue

≤0.005%

refractive index

n20/D 1.586 (lit.)

bp

184 °C (lit.)

mp

−6 °C (lit.)

solubility

water: soluble

density

1.022 g/mL at 25 °C (lit.)

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aniline is an aromatic primary amine that may be used as a reactant in the synthesis of organic intermediates such as 3-chloro-N-phenyl-pyridin-2-amine, (Z)-methyl 3-(phenylamino)but-2-enoate, 2-iodo-N-phenylbenzamide, 2,4-dichloroquinoline and N-(2-propynyl)aniline.

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Physical properties

Material darkens in storage to reddish-brown with no loss in purity.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Quinolines by Electrophilic Cyclization of N-(2-Alkynyl)Anilines: 3-Iodo-4-Phenylquinoline
Chen Y, et al
Organic Syntheses, 89, 294-306 (2012)
Synthesis of Tetrasubstituted 1H-Pyrazoles by Coppermediated
Coupling of Enaminones with Nitriles
Suri M and Glorius F
Organic Syntheses, 91, 211-220 (2014)
Synthesis of caboline
He l, et al
Organic Syntheses, 93, 272-285 (2016)
The application of fast scan cyclic voltammetry. Mechanistic study of the initial stage of electropolymerization of aniline in aqueous solutions.
Yang H and Bard AJ.
Journal of Electroanalytical Chemistry, 339(1), 423-449 (1992)
Potassium tert-Butoxide Mediated Synthesis of
Phenanthridinone
Bhakuni BS, et al
Organic Syntheses, 90, 164-173 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service