Skip to Content
Merck
All Photos(2)

Key Documents

32576

Supelco

Cimbuterol

VETRANAL®, analytical standard

Synonym(s):

2-Amino-5-(2-tert-butylamino-1-hydroxyethyl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N3O
CAS Number:
Molecular Weight:
233.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC(C)(C)NCC(O)c1ccc(N)c(c1)C#N

InChI

1S/C13H19N3O/c1-13(2,3)16-8-12(17)9-4-5-11(15)10(6-9)7-14/h4-6,12,16-17H,8,15H2,1-3H3

InChI key

YKKQAXQGZIBJFS-UHFFFAOYSA-N

General description

Cimbuterol is classified under the β-agonist group of compounds.

Application

Cimbuterol may be used as an analytical reference standard for the determination of the analyte in:
  • Pig liver, kidney and muscle samples by ultra-performance liquid chromatography-electrospray ionization-tandem mass spectrometry (UPLC-ESI-MS/MS).
  • Bovine and porcine urine, feed and hair samples by LC-ESI-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue analysis of beta-agonists in bovine and porcine urine, feed and hair using liquid chromatography electrospray ionisation tandem mass spectrometry.
Nielen MWF, et al.
Analytical and Bioanalytical Chemistry, 391(1), 199-210 (2008)
Multi-residual analysis of 16 ?-agonists in pig liver, kidney and muscle by ultra performance liquid chromatography tandem mass spectrometry.
Shao B, et al.
Food Chemistry, 114(3), 1115-1121 (2009)
Decheng Suo et al.
Journal of chromatography. A, 1526, 23-30 (2017-10-21)
A pseudo template molecularly imprinted polymer (MIP) for selective extraction of β-agonists was prepared using 4-hydroxyphenethyl alcohol as pseudo template molecule, methyl acrylic acid as functional monomer, and ethylene glycol dimethacrylate as cross-linker. Selectivity and adsorption capacity of prepared MIP
Kaylee R Mastrianni et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1084, 64-68 (2018-03-24)
An analytical procedure for the analysis of 10 β-adrenergic agonists (cimaterol, terbutaline, salbutamol, isoxsuprine, ractopamine, cimbuterol, clenbuterol, brombuterol, mabuterol and mapenterol) in pork meat was developed and validated using LC-MS/MS. An automated dispersive pipette extraction (DPX) was employed on a
Hongcheng Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1043, 176-186 (2016-08-21)
A simple, accurate and highly sensitive multiresidues method was developed for the determination of 9 anabolic steroids and 16 β-agonists in milk. Target compounds were extracted and separated by a modified QuEChERS (Quick, Easy, Cheap, Effective, Rugged, Safe) method with

Articles

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service