Skip to Content
Merck
All Photos(2)

Key Documents

53541

Supelco

Tulobuterol hydrochloride

VETRANAL®, analytical standard

Synonym(s):

2-tert-Butylamino-1-(2-chlorophenyl)ethanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClNO · HCl
CAS Number:
Molecular Weight:
264.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.CC(C)(C)NCC(O)c1ccccc1Cl

InChI

1S/C12H18ClNO.ClH/c1-12(2,3)14-8-11(15)9-6-4-5-7-10(9)13;/h4-7,11,14-15H,8H2,1-3H3;1H

InChI key

RSLNRVYIRDVHLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706019, T706030, T706020
Tulobuterol hydrochloride is a sympathomimetic amine, possessing bronchodilator activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tulobuterol hydrochloride may be used as a reference standard in determining the concentration of tulobuterol hydrochloride present in bovine liver homogenates and urine samples using gas chromatography coupled with tandem mass spectrometry (GC-MS-MS).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuwa Minami et al.
Internal medicine (Tokyo, Japan), 47(6), 503-509 (2008-03-18)
There is insufficient evidence for the efficacy of a transdermal tulobuterol patch (TP), although combination therapy with bronchodilators is recommended for chronic obstructive pulmonary disease (COPD). A randomized, controlled crossover study was conducted to evaluate the clinical efficacy and safety
Ki-Seok Kim et al.
Colloids and surfaces. B, Biointerfaces, 76(2), 404-409 (2009-12-26)
In this work, porous poly(epsilon-caprolactone) (PCL)/Eudragit RS 100 (ERS-100) microcapsules containing tulobuterol base as a model drug were prepared by a solvent evaporation method and the effect of the quaternary ammonium groups of ERS-100 on the release behaviors of the
Tetsuo Watanabe et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 131(10), 1483-1492 (2011-10-04)
The purpose of this study was to compare the transdermal transfer profiles of brand and generic tulobuterol patch formulations and to evaluate possible changes of in vivo kinetics resulting from increased transdermal transfer by means of pharmacokinetic analysis using reported
Chiyohiko Shindoh et al.
The Tohoku journal of experimental medicine, 218(4), 271-278 (2009-07-30)
Tulobuterol, a sympathomimetic drug used as a transdermal patch, increases normal diaphragm muscle strength. Because diaphragm muscle weakness (i.e. decrease of contraction) is a feature of bronchial asthma and sepsis, we examined the in vitro and in vivo effects of
Ki-Seok Kim et al.
Colloids and surfaces. B, Biointerfaces, 80(2), 240-246 (2010-07-14)
pH sensitive Pluronic F127 (PF127)/poly(acrylic acid) (PAAc) hydrogels containing tulobuterol-loaded silica were prepared with PF127 content. The effects of the PF127 content and silica on the sustained release behaviors of the PAAc hydrogels were investigated. The results showed that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service