Skip to Content
Merck
All Photos(1)

Key Documents

32409

Supelco

Nicarbazin

VETRANAL®, analytical standard

Synonym(s):

N,N′-bis(4-Nitrophenyl)urea compound with 4,6-dimethyl-2-pyrimidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N4O5 · C6H8N2O
CAS Number:
Molecular Weight:
426.38
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(C)nc(O)n1.[O-][N+](=O)c2ccc(NC(=O)Nc3ccc(cc3)[N+]([O-])=O)cc2

InChI

1S/C13H10N4O5.C6H8N2O/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22;1-4-3-5(2)8-6(9)7-4/h1-8H,(H2,14,15,18);3H,1-2H3,(H,7,8,9)

InChI key

UKHWDRMMMYWSFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicarbazin, an anti-coccidial drug, is an equimolar mixture of 4,4′-dinitrocarbanilide (DNC) and 2-hydroxy-4,6-dimethylpyrimidine (HDP). It is used for prophylactic treatment of coccidiosis in poultry birds.

Application

Nicarbazin may be used as a reference standard for the determination of nicarbazin residues in samples of chicken tissues, poultry feed, eggs and litter by high performance liquid chromatography (HPLC) method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

store under Argon

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface plasmon resonance biosensor screening of poultry liver and eggs for nicarbazin residues.
Mccarney B, et al.
Analytica Chimica Acta, 483(1-2), 165-169 (2003)
A new and rapid method for the determination of nicarbazin residues in poultry feed, eggs and muscle tissue using supercritical fluid extraction and high performance liquid chromatography.
Matabudul D, et al.
Analyst, 124(4), 499-502 (1999)
Micro high-performance liquid chromatography for the determination of nicarbazin in chicken tissues, eggs, poultry feed and litter.
Draisci R, et al.
Journal of Chromatography A, 697(1-2), 407-414 (1995)
Efficient HPLC method for the determination of nicarbazin, as dinitrocarbanilide in broiler liver.
Capurro E, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 822(1-2), 154-159 (2005)
G F Mathis et al.
Veterinary parasitology, 121(1-2), 1-9 (2004-04-28)
A 56-day floor pen study was conducted to determine the appropriate time to administer toltrazuril (Baycox) (TOL) for control of coccidiosis in broiler chickens. Litter was seeded with field strains of Eimeria acervulina, Eimeria maxima and Eimeria tenella. On Days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service