Skip to Content
Merck
All Photos(1)

Documents

31592

Supelco

Amprolium hydrochloride

VETRANAL®, analytical standard

Synonym(s):

1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H19N4Cl · HCl
CAS Number:
Molecular Weight:
315.24
Beilstein:
4118699
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

cationic

product line

VETRANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mol wt

315.24 g/mol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[Cl-].Cl[H].CCCc1ncc(C[n+]2ccccc2C)c(N)n1

InChI

1S/C14H19N4.2ClH/c1-3-6-13-16-9-12(14(15)17-13)10-18-8-5-4-7-11(18)2;;/h4-5,7-9H,3,6,10H2,1-2H3,(H2,15,16,17);2*1H/q+1;;/p-1

InChI key

PJBQYZZKGNOKNJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Wang et al.
Neuroscience, 144(3), 1045-1056 (2006-12-02)
Thiamine (vitamin B1) deficiency (TD) causes region selective neuronal loss in the brain; it has been used to model neurodegeneration that accompanies mild impairment of oxidative metabolism. The mechanisms for TD-induced neurodegeneration remain incompletely elucidated. Inhibition of protein glycosylation, perturbation
Sergiy Chornyy et al.
Acta biochimica Polonica, 54(2), 315-322 (2007-05-16)
Recent evidence suggests that alterations in oxidative metabolism induced by thiamine deficiency lead to neuronal cell death. However, the molecular mechanisms underlying this process are still under extensive investigation. Here, we report that rat pheochromocytoma PC-12 cells differentiated in the
Gabrielle Young et al.
Veterinary parasitology, 178(3-4), 346-349 (2011-02-22)
This study evaluated the efficacy of two different doses of amprolium in goats heavily infected with pathogenic Eimeria species. Forty Boer goat kids ranging from 3 to 5 months of age with naturally occurring coccidiosis were randomly divided into 2
Clara Lemos et al.
European journal of pharmacology, 682(1-3), 37-42 (2012-03-06)
The aim of this study was to characterize the intestinal absorption of thiamine, by investigating the hypothesis of an involvement of Organic Cation Transporter (OCT) family members in this process. [(3)H]-T(+) uptake was found to be: 1) time-dependent, 2) Na(+)-
D Bizon-Zygmańska et al.
Neurochemistry international, 59(2), 208-216 (2011-06-16)
Inhibition of pyruvate (PDHC) and ketoglutarate (KDHC) dehydrogenase complexes induced by thiamine pyrophosphate deficits is known cause of disturbances of cholinergic transmission in the brain, yielding clinical symptoms of cognitive, vegetative and motor deficits. However, particular alterations in distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service