Skip to Content
Merck
All Photos(4)

Key Documents

M5273

Sigma-Aldrich

Monensin sodium salt

90-95% (TLC)

Synonym(s):

Monensin A sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H61NaO11
CAS Number:
Molecular Weight:
692.85
Beilstein:
4122200
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

90-95% (TLC)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
parasites
viruses

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[H][C@]1([C@@]2(CC)O[C@]([C@@]3(C)OC4(C[C@H](O)[C@@H](C)[C@@]([H])([C@@H](C)[C@@H](OC)[C@@H](C([O-])=O)C)O4)CC3)([H])CC2)[C@@H](C)C[C@]([H])([C@@]5([H])O[C@@](O)(CO)[C@H](C)C[C@@H]5C)O1.[Na+]

InChI

1S/C36H62O11.Na/c1-10-34(31-20(3)16-26(43-31)28-19(2)15-21(4)36(41,18-37)46-28)12-11-27(44-34)33(8)13-14-35(47-33)17-25(38)22(5)30(45-35)23(6)29(42-9)24(7)32(39)40;/h19-31,37-38,41H,10-18H2,1-9H3,(H,39,40);/q;+1/p-1/t19-,20-,21+,22+,23-,24-,25-,26+,27+,28-,29+,30-,31+,33-,34-,35?,36-;/m0./s1

InChI key

XOIQMTLWECTKJL-BEMBKCOJSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chemical structure: polyether
Monensin is a polyether ionophoric antibiotic, which is produced by Streptomyces cinnamonensis.[1] It is used to treat bacterial, fungal and parasitic infections. Monensin prevents the growth of colon cancer cells.[2] It facilitates the transport of sodium and potassium ions between intracellular and extracellular spaces.[3] Monensin prevents coccidiosis in poultry production.[3]

Application

Monensin sodium salt has been used:
  • for the intracellular staining of cytokines[4][5]
  • to study its effects on the regulation of nuclear factor-κ B (NF-κB ) activation[6]
  • as a positive control to study its effects on in vitro rumen fermentation[7][8]

Biochem/physiol Actions

Na+ ionophore; blocks glycoprotein secretion; may induce catecholamine secretion from chromaffin cells. Useful in potentiometric and spectroscopic studies of alkali metal ion complexes.

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - STOT RE 2

Target Organs

Heart,muscle

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and antimicrobial properties of monensin A and its derivatives: summary of the achievements
Lowicki D and Huczynski A
BioMed Research International, 2013 (2013)
Gfi1, a transcriptional repressor, inhibits the induction of the T helper type 1 programme in activated CD 4 T cells
Suzuki J, et al.
Immunology, 147(4), 476-487 (2016)
In vitro screening of essential oil active compounds for manipulation of rumen fermentation and methane mitigation
Joch M, et al.
Asian-Australasian Journal of Animal Sciences, 29(7), 952-952 (2016)
Wntbeta catenin-mediated signaling commonly altered in colorectal cancer
Progress in Molecular Biology and Translational Science, 144, 49-68 (2016)
Golgi tethering factor golgin-97 suppresses breast cancer cell invasiveness by modulating NF-κB activity
Hsu RM, et al.
Cell communication and signaling : CCS, 16(1), 19-19 (2018)

Questions

  1. How long is this stable as a 1mg/ml stock solution in ethanol when stored at 4ºC?

    1 answer
    1. Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service