Skip to Content
Merck

303003

Hexane-d14

99 atom % D

Synonym(s):

Tetradecadeuterohexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CD3(CD2)4CD3
CAS Number:
Molecular Weight:
100.26
UNSPSC Code:
12142201
NACRES:
NA.21
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1841418
Isotopic purity:
99 atom % D
Assay:
99% (CP)
Mass shift:
M+14
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3 (vs air)

Quality Segment

vapor pressure

132 mmHg ( 20 °C)

isotopic purity

99 atom % D

assay

99% (CP)

form

liquid

expl. lim.

7.7 %

technique(s)

NMR: suitable

impurities

≤0.5% water
water

refractive index

n20/D 1.3718 (lit.)

bp

69 °C

mp

-95 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

mass shift

M+14

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2

InChI key

VLKZOEOYAKHREP-ZLKPZJALSA-N

General description

Achieve sharper, more reliable nuclear magnetic resonance (NMR) spectra with our comprehensive range of deuterated solvents. Each solvent delivers exceptional chemical purity and the highest isotopic enrichment, ensuring a stable deuterium lock signal, minimal residual proton signal interference, and a reliable chemical shift reference for every experiment.



Ultra-low residual water content - verified by both Karl Fischer titration and NMR methods - supports moisture-sensitive applications, while rigorous batch-to-batch quality control delivers consistent, reproducible performance you can trust in research and quality control laboratories.


Our portfolio spans the most widely used NMR solvents available in multiple deuteration levels and purity grades to match the exact requirements of your analysis. Available in a variety of safe, flexible formats - including glass bottles, glass ampules, serum bottles, and our exclusive moisture-protective Sure/Seal™ bottles - to suit every lab′s handling and storage needs.

Application

Deuterated solvents are indispensable wherever chemical research is carried out, supporting nuclear magnetic resonance (NMR) spectroscopy - the leading method for structural elucidation of organic molecules. As a non-destructive, information-rich technique, NMR reveals atomic connectivity within a molecule, its spatial orientation, and its motion in its natural environment, offering a comprehensive picture of molecular structure and dynamics.

Other Notes

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

ChemisTwin is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC


Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

WGK

WGK 3

Flash Point (°F)

10.4 °F - closed cup

Flash Point (°C)

-12 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library