Skip to Content
Merck
All Photos(2)

Key Documents

494526

Sigma-Aldrich

Heptane

greener alternative

biotech. grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
biotech. grade
Assay:
≥99%
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

grade

biotech. grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

<0.005% water

evapn. residue

<0.0002%

color

APHA: ≤10

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

solubility

acetone: miscible(lit.)
carbon tetrachloride: soluble(lit.)
ethanol: very soluble(lit.)
water: insoluble(lit.)

density

0.684 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 197 nm Amax: 1.00
λ: 210 nm Amax: 0.40
λ: 220 nm Amax: 0.15
λ: 230 nm Amax: 0.05
λ: 260-400 nm Amax: 0.01

greener alternative category

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptane (n-Heptane) is an alkane hydrocarbon. It is a primary reference fuel (PRF) employed for determining the octane number of fuels in internal combustion engines. Its cetane number and octane number has been reported to be 56 and 0 respectively. Its thermal decomposition has been studied under different conditions of temperature and pressure. Its oxidative dehydrogenation in the presence of magnesium oxide supported vanadium catalyst has been investigated. Molybdenum phosphide supported on Hβ zeolite (MoP/Hβ) catalyzed hydroisomerization of n-heptane has been reported to be enhanced by doping with secondary metals (Cr, Ni or Ce).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information. Heptane is a linear hydrocarbon that is considered as a greener alternative to the more toxic hexane.

Application

Greener alternatives to Hexane.
Heptane (n-Heptane) may be used:
  • To compose a solvent mixture with diethyl ether for the purification of N-acylpyrrolidine derivatives of fatty acid methyl esters (FAMEs) by TLC.
  • To generate of chiral alcohols and heptanones, via whole cell double oxidation.
  • As a solvent in the synthesis of isopropyl acetate, via esterification of acetic acid with isopropyl alcohol catalyzed by lipase immobilized on silica.

Features and Benefits

Greener alternative for hexane

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High pressure pyrolysis of n-heptane.
Chakraborty JP and Kunzru D.
Journal of Analytical and Applied Pyrolysis, 86(1), 44-52 (2009)
Hydroisomerization of n-heptane over MoP/Hβ catalyst doped with metal additive.
Liu P, et al.
Fuel Processing Technology, 131, 311-316 (2015)
Whole-cell double oxidation of n-heptane.
Muller CA, et al.
Journal of Biotechnology, 191, 196-204 (2014)
Activation of n-Heptane: A Study with VMgO Catalysts.
Dasireddy VDBC, et al.
Catalysis Letters, 144(4), 590-597 (2014)
Madan Lal Verma et al.
Enzyme research, 2011, 919386-919386 (2011-05-24)
Selective production of fragrance fatty acid ester from isopropanol and acetic acid has been achieved using silica-immobilized lipase of Bacillus cereus MTCC 8372. A purified thermoalkalophilic extracellular lipase was immobilized by adsorption onto the silica. The effects of various parameters

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service