Skip to Content
Merck
All Photos(1)

Documents

248266

Sigma-Aldrich

Xylidyl blue I

synthetic colorimetric reagent

Synonym(s):

4-Hydroxy-3-[2-hydroxy-3-(2,4-dimethylphenylaminocarbonyl)-1-naphthylazobenzenesulfonic acid sodium salt, Magonsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3[N=NC10H5(OH)[CONHC6H3(CH3)2]]SO3Na
CAS Number:
Molecular Weight:
513.50
Beilstein:
4107363
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

form

powder or crystals

technique(s)

titration: suitable

color

dark violet

λmax

612 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1ccc(NC(=O)c2cc3ccccc3c(\N=N\c4cc(ccc4O)S([O-])(=O)=O)c2O)c(C)c1

InChI

1S/C25H21N3O6S.Na/c1-14-7-9-20(15(2)11-14)26-25(31)19-12-16-5-3-4-6-18(16)23(24(19)30)28-27-21-13-17(35(32,33)34)8-10-22(21)29;/h3-13,29-30H,1-2H3,(H,26,31)(H,32,33,34);/q;+1/p-1/b28-27+;

InChI key

LKSHJHJGLORTGD-RXQWRGDBSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Xylydil Blue I is a synthetic colorimetric reagent that has been widely used for the spectrophotometric detection of magnesium.

Application

Xylydil Blue I is suitable to determine serum magnesium levels using Mg- Xylydil Blue I complex based method.

Biochem/physiol Actions

Xylydil Blue I forms a purple complex with the magnesium ion in alkaline conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Measurement of magnesium in the human sera with xylidyl-blue I. I. The effect of phosphate ion].
Y Takubo et al.
Rinsho byori. The Japanese journal of clinical pathology, 31(11), 1244-1248 (1983-11-01)
M J Stone et al.
Clinical chemistry, 42(9), 1474-1477 (1996-09-01)
We have evaluated an enzymatic assay for the determination of Mg in serum, based on the activation of a chemically modified form of isocitrate dehydrogenase by Mg2+. In the presence of potassium isocitrate and NADP+, NADPH, which absorbs light at
Measurement of magnesium in serum and urine with a random access analyzer by use of a modified xylidyl blue-1 procedure.
D Ratge et al.
Clinica chimica acta; international journal of clinical chemistry, 159(2), 197-203 (1986-09-15)
M R Wills et al.
Magnesium, 5(5-6), 317-327 (1986-01-01)
A variety of methods have been reported for the quantitative analysis of magnesium in serum and other biological fluids. The methods may be classified into the following groups: magnesium ammonium phosphate; 8-hydroxyquinoline; titan yellow; dihydroxyazobenzene using fluorimetry; Schwarzenbach ethylenediaminetetraacetic acid;
Determination of urine magnesium using the magon dye method on the 'Monarch' centrifugal analyser.
W Davidson et al.
Annals of clinical biochemistry, 27 ( Pt 6), 595-596 (1990-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service