All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H7NO4S · xH2O
CAS Number:
Molecular Weight:
225.22 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
>300 °C (lit.)
SMILES string
O.Oc1ccc(c2cccnc12)S(O)(=O)=O
InChI
1S/C9H7NO4S.H2O/c11-7-3-4-8(15(12,13)14)6-2-1-5-10-9(6)7;/h1-5,11H,(H,12,13,14);1H2
InChI key
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
Application
Pre- and post-column reagent for fluorescence detection of metal ions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of Chromatography A, 671, 121-121 (1994)
Chem. Abstr., 117, 103167y-103167y (1992)
Fenxi Huaxue, 20, 638-638 (1992)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service