Skip to Content
Merck
All Photos(3)

Documents

21198

Supelco

Calcium ionophore IV

Selectophore, function tested

Synonym(s):

N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide, N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide, ETH 5234

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H100N2O3
CAS Number:
Molecular Weight:
801.36
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

mp

50-53 °C

SMILES string

CCCCCCCCCCCCCCCCCCN(CCCCCCCCCCCCCCCCCC)C(=O)COCC(=O)N(C1CCCCC1)C2CCCCC2

InChI

1S/C52H100N2O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-39-45-53(46-40-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)51(55)47-57-48-52(56)54(49-41-35-33-36-42-49)50-43-37-34-38-44-50/h49-50H,3-48H2,1-2H3

InChI key

CLLKVTAGSOWKHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Calcium ionophore IV (ETH 5234) was used in preparing membrane for the pseudoreference electrode in tetrahydrofuran, in a study to demonstrate electrochemical sample matrix elimination for trace level potentiometric detection.
Calcium ionophore IV may be used in the preparation of Ca2+-selective membrane utilizing AlGaN/GaN transistors for potentiometric calcium ion detection.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Extremely lipophilic Ca2+-selective ionophore for chemical sensors of long lifetime; it induces selectivities for Ca over Mg, Na and K of 104, 106, and 108, respectively.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Y Chumbimuni-Torres et al.
Analytical chemistry, 80(15), 6114-6118 (2008-06-24)
Potentiometric sensors are today sufficiently well understood and optimized to reach ultratrace level (subnanomolar) detection limits for numerous ions. In many cases of practical relevance, however, a high electrolyte background hampers the attainable detection limits. A particularly difficult sample matrix
Mohsen Asadnia et al.
Analytica chimica acta, 987, 105-110 (2017-09-17)
We demonstrate highly selective and sensitive potentiometric ion sensors for calcium ion detection, operated without the use of a reference electrode. The sensors consist of AlGaN/GaN heterostructure-based transistor devices with chemical functionalisation of the gate area using poly (vinylchloride)-based (PVC)
Jingwei Zhu et al.
Analytical chemistry, 82(1), 436-440 (2009-12-04)
A novel magnesium-selective ion-channel mimetic sensor based on a well-known calcium ionophore, ETH 5234, was demonstrated. Due to different stoichiometry of the ionophore with Mg(2+) and Ca(2+), the sensor exhibits high selectivity to Mg(2+) by controlling ionophore concentration on the
Lowering the Detection Limit of Solvent Polymeric Ion-Selective Membrane Electrodes. 2. Influence of Composition of Sample and Internal Electrolyte Solution
Sokalski, T.; et al.
Analytical Chemistry, 71(6), 1210-1214 (1999)
P. Gehrig et al.
Chimia, 43, 377-377 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service