Skip to Content
Merck
All Photos(2)

Documents

00744

Supelco

Magnesium ionophore VII

Selectophore, function tested

Synonym(s):

4,13-[Bis(N-adamantylcarbamoyl)acetyl]-1,7,10,16,tetraoxa-4,13-diazacyclooctadecane, K22B5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H60N4O8
CAS Number:
Molecular Weight:
700.91
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

mp

85-90 °C (lit.)

storage temp.

−20°C

SMILES string

O=C(CC(=O)N1CCOCCOCCN(CCOCCOCC1)C(=O)CC(=O)NC23CC4CC(CC(C4)C2)C3)NC56CC7CC(CC(C7)C5)C6

InChI

1S/C38H60N4O8/c43-33(39-37-21-27-13-28(22-37)15-29(14-27)23-37)19-35(45)41-1-5-47-9-10-49-7-3-42(4-8-50-12-11-48-6-2-41)36(46)20-34(44)40-38-24-30-16-31(25-38)18-32(17-30)26-38/h27-32H,1-26H2,(H,39,43)(H,40,44)

InChI key

IPVOJCDBVJUPRP-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Other Notes

Mg-ionophore with excellent selectivity

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L Lvova et al.
Analytica chimica acta, 988, 96-103 (2017-09-17)
A systematic study of a series of diaza-18-crown-6 8-hydroxyquinoline (DCHQ) chemosensors, devoted to Mg(II) ion detection, was performed. Functionalization of DCHQ by peripheral substituents allowed the development of novel all-solid-state optodes via inclusion inside PVC-based polymeric films. The influence on
Design and Synthesis of Calcium and Magnesium Ionophores Based on Double-Armed Diazacrown Ether Compounds and Their Application to an Ion-Sensing Component for an Ion-Selective Electrode
Suzuki, K., et al.
Analytical Chemistry, 67, 324-324 (1995)
A Comparison of Neutral Mg2+-Selective Ionophores in Solvents Polymeric Membranes: Complex Stoichiometry
Zhang, W., et al.
Analytical Sciences, 16, 11-18 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service