Skip to Content
Merck
All Photos(3)

Documents

21192

Supelco

Calcium ionophore I

Selectophore, function tested, ≥99.0%

Synonym(s):

(−)-(R,R)-N,N′-Bis-[11-(ethoxycarbonyl)undecyl]-N,N′,4,5-tetramethyl-3,6-dioxaoctane-diamide, Diethyl N,N′-[(4R,5R)-4,5-dimethyl-1,8-dioxo-3,6-dioxaoctamethylene]bis(12-methylaminododecanoate), ETH 1001

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O8
CAS Number:
Molecular Weight:
684.99
Beilstein:
2343275
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (HPLC)
≥99.0%

quality

function tested

SMILES string

CCOC(=O)CCCCCCCCCCCN(C)C(=O)CO[C@H](C)[C@@H](C)OCC(=O)N(C)CCCCCCCCCCCC(=O)OCC

InChI

1S/C38H72N2O8/c1-7-45-37(43)27-23-19-15-11-9-13-17-21-25-29-39(5)35(41)31-47-33(3)34(4)48-32-36(42)40(6)30-26-22-18-14-10-12-16-20-24-28-38(44)46-8-2/h33-34H,7-32H2,1-6H3/t33-,34-/m1/s1

InChI key

CJAONIOAQZUHPN-KKLWWLSJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Calcium ionophore I (ETH 1001) was used in an experimental study, conducted to determine the intracellular and extracellular changes of calcium ions in hypoxia and ischemia in rat brain.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Used as ionophore in membrane electrodes for the determination of Ca2+-activities; Useful for transport studies in biological membranes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rita I Jabr et al.
Pflugers Archiv : European journal of physiology, 468(11-12), 1945-1955 (2016-10-21)
Cardiac arrhythmias are associated with raised intracellular [Ca2+] and slowed action potential conduction caused by reduced gap junction (GJ) electrical conductance (Gj). Ventricular GJs are composed of connexin proteins (Cx43), with Gj determined by Cx43 phosphorylation status. Connexin phosphorylation is
Anna Kisiel et al.
Talanta, 220, 121358-121358 (2020-09-16)
An alternative optical signal transduction mechanism for ion-selective optodes is proposed. The nanostructural sensors benefit from ion-selective reversing aggregation caused quenching yielding turn-on, bright and highly stable optical signals. Selective incorporation of analyte results in transformation of the polymer dye
I A Silver et al.
The Journal of general physiology, 95(5), 837-866 (1990-05-01)
Changes in intra- and extracellular free calcium concentration were evaluated with ion-selective microelectrodes during periods of anoxia and ischemia in three different regions of intact rat brain. Recordings stable for at least 2 min and in most cases for 4-6
W. Simon et al.
Annals of the New York Academy of Sciences, 307, 52-52 (1978)
N Abuaf et al.
Journal of allergy, 2012, 580873-580873 (2011-12-22)
Background. An in vitro basophil activation test, based on the detection of CD63 upregulation induced by NSAIDs, has been described. Its clinical significance remains controversial. Objectives. In patients with a history of nonallergic NSAID hypersensitivity, stratified according to the severity

Articles

Sigma-Aldrich.com presents an article on Selectophore Ionophores

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service