Skip to Content
Merck
All Photos(2)

Documents

20909

Supelco

Cadmium ionophore I

Selectophore

Synonym(s):

N,N,N′,N′-Tetrabutyl-3,6-dioxaoctanedi(thioamide), ETH 1062

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H44N2O2S2
CAS Number:
Molecular Weight:
432.73
Beilstein:
4521607
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

mp

34-36 °C

SMILES string

CCCCN(CCCC)C(=S)COCCOCC(=S)N(CCCC)CCCC

InChI

1S/C22H44N2O2S2/c1-5-9-13-23(14-10-6-2)21(27)19-25-17-18-26-20-22(28)24(15-11-7-3)16-12-8-4/h5-20H2,1-4H3

InChI key

NKWPVGBRWCCVGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Selective ionophore for cadmium used in solvent polymeric membranes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haipeng Guo et al.
Physiologia plantarum, 159(3), 340-353 (2016-10-28)
Miscanthus is a vigorous perennial Gramineae genus grown throughout the world as a promising bioenergy crop and generally regarded as heavy metal tolerant due to its ability to absorb heavy metals. However, little is known about the mechanism for heavy
Hua Xu et al.
Chemosphere, 190, 154-165 (2017-10-11)
Cadmium (Cd) is a detrimental environmental pollutant. Duckweeds have been considered promising candidates for Cd phytoremediation. Although many physiological studies have been conducted, the molecular mechanisms underlying Cd hyperaccumulation in duckweeds are largely unknown. In this study, clone 6001 of
Lian-Zhen Li et al.
Environmental toxicology and chemistry, 36(4), 1038-1046 (2016-10-30)
Uptake and membrane transport of cadmium (Cd) in roots of the hyperaccumulator Sedum plumbizincicola X.H. Guo et S.B. Zhou ex L.H. Wu was characterized by assessing the impact of various inhibitors and ion channel blockers on Cd accumulation as well
Jie Ma et al.
Aquatic toxicology (Amsterdam, Netherlands), 202, 136-144 (2018-07-22)
The nitrogen (N) and phosphorus (P) loadings in coastal waters have greatly increased due to anthropogenic input. However, the silicate levels have remained stable in the past decades, leading to nutrient status alternating from N limitation to Si limitation. Few
Xiaohui Chen et al.
Journal of experimental botany, 69(21), 5279-5291 (2018-08-14)
To develop elite crops with low cadmium (Cd), a fundamental understanding of the mechanism of Cd uptake by crop roots is necessary. Here, a new mechanism for Cd2+ entry into rice root cells was investigated. The results showed that Cd2+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service