Skip to Content
Merck
All Photos(1)

Documents

21102

Sigma-Aldrich

Calcium chloride dihydrate

tested according to Ph. Eur.

Synonym(s):

Calcii chloridum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

mp

176 °C (dec.) (lit.)

solubility

water: soluble 147 g/L at 20 °C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium chloride dihydrate has been used for synthesis of chemical hydrogels based on telechelic Poly (vinyl alcohol) (PVA). It has also been used for investigating the morphology of polystyrene-based emulsion-templated (PolyHIPE) materials.

Biochem/physiol Actions

Calcium chloride act as precipitants during protein crystallization process. It also enhances the competence of Escherichia coli cells as well as strengthened β-lactoglobulin gels by forming crossbridges.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaio Paradossi et al.
Journal of materials science. Materials in medicine, 14(8), 687-691 (2004-09-07)
In this paper, we present some new case examples where the chemical versatility of poly (vinyl alcohol) (PVA) can be used for potential biomedical applications. PVA, the polymeric material used for designing new nanostructured devices, is water soluble, biocompatible and
Tailoring the morphology of emulsion-templated porous polymers.
Carnachan, Ross J., et al
Soft Matter, 2, 608-616 (2006)
Gelation of beta-Lactoglobulin: Effects of Sodium Chloride and Calcium Chloride on the Rheological and Structural Properties of Gels.
D.M. Mulvihill
Journal of Food Science, 53, 231-236 (1988)
Christian Epple et al.
Biomaterials, 192, 118-127 (2018-11-19)
Large and complex bone defects represent challenging clinical scenarios, typically requiring autologous vascularized bone transplants. In order to bypass the numerous associated limitations, here we aimed at ectopically prefabricating a bone graft surrogate with vascular pedicle. A hollow cylinder of
Matthew S Kimber et al.
Proteins, 51(4), 562-568 (2003-06-05)
Protein crystallization is a major bottleneck in protein X-ray crystallography, the workhorse of most structural proteomics projects. Because the principles that govern protein crystallization are too poorly understood to allow them to be used in a strongly predictive sense, the

Related Content

Find solutions for bulk production, custom packaging, and personalized chemical services to meet your unique research, development, and manufacturing requirements in your industries, including pharmaceuticals, biotechnology, and research institutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service