Skip to Content
Merck
All Photos(5)

Key Documents

Z560030

Levgo® smartSpatula® disposable PP laboratory spatula, Look for the V at the neck

L 210 mm, blue, standard

Synonym(s):

disposable spatula, microspatula, spatula

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.35
Pricing and availability is not currently available.

material

polypropylene

packaging

pack of 300 ea
case of 7500 ea

manufacturer/tradename

LevGo Inc 17211

L

210 mm

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Versatile Double-ended polypropylene tool that can scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of chemicals and biological materials.


LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.

Features and Benefits

  • The V design at the neck of the disposable lab spatula is a trademark of LevGo, Inc.
  • No Searching, washing or contamination.
  • 100% Polypropylene is autoclavable and resistant to dilute acids and bases.
  • Polypropylene will not shatter after exposure to liquid nitrogen frozen samples.
  • Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer.
  • SGS-Tested for US and EU food contact standards.
  • Sustainability – LevGo plants a tree for every case sold.

Legal Information

US Patent 8,709,362
LevGo is a registered trademark of LevGo Inc.
Look for the V at the neck is a trademark of LevGo Inc.
smartSpatula is a registered trademark of LevGo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service