Skip to Content
Merck
All Photos(1)

Key Documents

46385

Supelco

Florisil® Inorganic Adsorbent

60-100 mesh

Synonym(s):

Florisil®, Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Florisil® Adsorbent for Chromatography, 60-100 mesh

grade

standard

Quality Level

form

coarse powder

technique(s)

LPLC: suitable

matrix

Magnesium silicate hydrate

matrix active group

magnesium silicate hydrate

particle size

149-250 μm
60-100 mesh

pore size

60 Å mean pore size

separation technique

hydrophilic interaction (HILIC)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Florisil, adsorbent for the separation of lipids, is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate.[1] Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers.[2] Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.[3]

Application

Florisil was used:
  • as column packing material in column chromatography[4]
  • as an adsorbent in gas chromatography with electron capture detector (GC/ECD)[5], capillary column chromatography[6], adsorption column chromatography[7]
  • in clean-up procedures in gas chromatography-tandem mass spectrometry[7] and GC-FID[8]

Caution

may contain some dark particles

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved gas chromatographic method for the determination of perfluorocarbon tracers in the atmosphere
Lagomarsino, Raymond J.
Journal of Chromatography A, 34, 405-412 (1996)
Andrew C Singer et al.
Environmental toxicology and chemistry, 22(9), 1998-2004 (2003-09-10)
This study investigated the interactive effects of bioaugmentation, biostimulation, and the rhizosphere during remediation of Aroclor 1242-contaminated soil. Treatments were repeatedly augmented with polychlorinated bipheny (PCB)-degrading bacteria, inducers (carvone and salicylic acid), surfactant (sorbitan trioleate), minimal salts medium in a
Determination of atmospheric perfluorocarbon background concentrations of fL/L range at the western coastal area of Korea
Kim, HyeKyeong, et al.
Atmospheric Environment, 1, 4-4 (2002)
R C Prados-Rosales et al.
Journal of chromatography. A, 993(1-2), 121-129 (2003-05-09)
A rapid analytical method for determination of organochlorine pesticide residues in sunflower seeds based on focused microwave-assisted Soxhlet extraction has been developed. The main factors affecting the extraction efficiency--namely microwave power, irradiation time, volume of extractant and number of cycles--were
The covalently bound flavin of Chromatium cytochrome c552. 1. Evidence for cysteine thiohemiacetal at the 8 alpha position.
W H Walker et al.
European journal of biochemistry, 48(2), 439-448 (1974-10-02)

Related Content

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service