Skip to Content
Merck
All Photos(2)

Key Documents

B9757

Sigma-Aldrich

Betulin

≥98%

Synonym(s):

Betulinic alcohol, Betulinol, Betulol, Lup-20(29)-ene-3β,28-diol, Trochol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

powder

mp

256-257 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3([H])[C@@]4(C)CC[C@H](O)C(C)(C)[C@]4([H])CC[C@@]3(C)[C@]1(C)CC[C@@]5(CO)CC[C@@H](C(C)=C)[C@]25[H]

InChI

1S/C30H50O2/c1-19(2)20-10-15-30(18-31)17-16-28(6)21(25(20)30)8-9-23-27(5)13-12-24(32)26(3,4)22(27)11-14-29(23,28)7/h20-25,31-32H,1,8-18H2,2-7H3/t20-,21+,22-,23+,24-,25+,27-,28+,29+,30+/m0/s1

InChI key

FVWJYYTZTCVBKE-ROUWMTJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betulin, a triterpene with lupane nucleus,[1] is found in the bark of birch trees.[2] It undergoes rearrangement in the presence of acid agents to form allobetulin.[3]

Application

Betulin may be used in the preparation of betulinic acid,[4] which shows anti-HIV, antimalarial and anti-inflammatory activities.[2]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy Horwedel et al.
Journal of medicinal chemistry, 53(13), 4842-4848 (2010-06-10)
A novel approach to circumvent multidrug resistance is hybridization of natural products in dimers. We analyzed homodimers of two artesunic acid molecules and heterohybrids of artesunic acid and betulin in human CCRF-CEM and multidrug-resistant P-glycoprotein-overexpressing CEM/ADR5000 leukemia cells. Multidrug-resistant cells
Betulin and its derivatives. Chemistry and biological activity.
Tolstikov GA, et al.
Chemistry for Sustainable Development, 13, 1-29 (2005)
A concise semi-synthetic approach to betulinic acid from betulin.
Kim DSHL, et al.
Synthetic Communications, 27(9), 1607-1612 (1997)
A bicentennial of betulin.
Hayek EWH, et al.
Phytochemistry, 28(9), 2229-2242 (1989)
Milan Urban et al.
Bioorganic & medicinal chemistry, 20(11), 3666-3674 (2012-05-04)
The aim of this work was to synthesize a set of heterocyclic derivatives of lupane, lup-20(29)-ene, and 18α-oleanane, and to investigate their cytotoxic activities. Some of those heterocycles were previously known in the oleanane (allobetulin) group; however, to our knowledge

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service