Skip to Content
Merck
All Photos(1)

Key Documents

53017

Supelco

α-Amyrin

analytical standard

Synonym(s):

Urs-12-en-3β-ol, Viminalol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Beilstein:
1916550
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@H]1CC[C@]2(C)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C

InChI

1S/C30H50O/c1-19-11-14-27(5)17-18-29(7)21(25(27)20(19)2)9-10-23-28(6)15-13-24(31)26(3,4)22(28)12-16-30(23,29)8/h9,19-20,22-25,31H,10-18H2,1-8H3/t19-,20+,22+,23-,24+,25+,27-,28+,29-,30-/m1/s1

InChI key

FSLPMRQHCOLESF-SFMCKYFRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Reference material for the analysis of latex milk

Biochem/physiol Actions

Triterpene that has broad-spectrum analgesic properties for which the underlying mechanism is poorly understood but which may involve the inhibition of protein kinase A (PKA).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flávia Almeida Santos et al.
Lipids in health and disease, 11, 98-98 (2012-08-08)
Pentacyclic triterpenes in general exert beneficial effects in metabolic disorders. This study investigated the effects of α, β-amyrin, a pentacyclic triterpene mixture from the resin of Protium heptaphyllum on blood sugar level and lipid profile in normal and streptozotocin (STZ)-induced
Javid Hussain et al.
Journal of Asian natural products research, 14(1), 22-26 (2012-01-24)
Two new phthalates bis(2-ethylundecyl)phthalate and bis(2-ethyltridecyl)phthalate have been isolated from the chloroform-soluble portion of the whole plant of Nepeta clarkei along with one known compound β-amyrin. The structures of the two new compounds and β-amyrin were assigned on the basis
Vanessa Janovik et al.
Natural product research, 26(1), 66-71 (2011-08-11)
The Lecythidaceae family is composed of 25 genera and 400 species that occur in the form of trees with a pantropical distribution. The genera Cariniana belongs to the family Lecythidaceae, and there are few reports considering these species. In this
Edyta Biskup et al.
Acta biochimica Polonica, 59(2), 255-260 (2012-05-12)
Rhaponticum carthamoides plants ("maral root") are widely used in Siberian folk medicine. The present study reports for the first time the presence of pentacyclic terpenoid, α-amyrin, in methanol extract from leaves of this plant. α-Amyrin induced proliferation of human keratinocytes
A C Siani et al.
Phytochemical analysis : PCA, 23(5), 529-539 (2012-02-11)
Non-volatile oleoresins from neotropical Burseraceae are traditionally used for craft, technological and medicinal purposes. The crude resin is usually sold in popular markets of the forest communities. Adding value to this rainforest raw material requires establishing its composition. To analyse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service