Skip to Content
Merck
All Photos(1)

Documents

02390

Sigma-Aldrich

1,2-Ethanedithiol

≥98.0% (GC)

Synonym(s):

1,2-Dimercaptoethane, Dithioglycol, Ethylene mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2SH
CAS Number:
Molecular Weight:
94.20
Beilstein:
505946
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

4.8 mmHg ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.558 (lit.)
n20/D 1.558

bp

144-146 °C (lit.)

mp

−41 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

SMILES string

SCCS

InChI

1S/C2H6S2/c3-1-2-4/h3-4H,1-2H2

InChI key

VYMPLPIFKRHAAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Ethanedithiol (EDT) is an organosulfur compound that is used as an organic building block in chemical synthesis. It is also used as a ligand for metal ions to prepare metal complexes.

Application

1,2-Ethanedithiol can be used to synthesize:      
  • 1,3-Dithiolanes by reacting with aldehydes or ketones.
  • Dithiazepan-3-yl-alkanoic acids by cyclocondensation reaction with amino acids and formaldehyde.
  • EDT can also be used as a scavenger in solid phase peptide synthesis for the efficient deprotection of the peptide resin.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Majed Halwani et al.
International journal of pharmaceutics, 373(1-2), 141-146 (2009-05-12)
Recurrent pulmonary infection and inflammation are major risk factors for high morbidity and mortality in patients with cystic fibrosis (CF). As such, frequent antibiotic use and drug resistant bacterial strains are main concerns in individuals with CF. Bacterial virulence and
Eunice F S Vieira et al.
Journal of colloid and interface science, 289(1), 42-47 (2005-07-13)
The nature of interactions between metal ions Ag(I), Hg(II), Cu(II) and chitosan derivative of 1,2-ethanedithiol, QTDT, was investigated by isothermal calorimetry using the membrane breaking technique. Simultaneous determination of thermal effects, Q(int), and amount of cation that interacts, n(int), are
Benjamin J Miller et al.
The journal of physical chemistry. A, 114(48), 12692-12700 (2010-11-12)
Vibrational spectra of vapor-phase 1,2-ethanedithiol and 2-mercaptoethanol were recorded to investigate weak intramolecular interactions. The spectra were recorded with conventional absorption spectroscopy and laser photoacoustic spectroscopy in the 2000-11,000 cm(-1) region. The room temperature spectra of each molecule are complicated
Misagh Alipour et al.
The Journal of antimicrobial chemotherapy, 65(4), 684-693 (2010-02-18)
This study examined the activities of tobramycin and bismuth against quorum sensing, virulence factors and biofilms of Pseudomonas aeruginosa by co-encapsulating the agents in liposomes in order to achieve greater delivery of the agents. The inhibitory effects of the agents
Efficient heterojunction photovoltaic cell utilizing nanocomposites of lead sulfide nanocrystals and a low-bandgap polymer.
Jangwon Seo et al.
Advanced materials (Deerfield Beach, Fla.), 23(34), 3984-3988 (2011-07-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service