Skip to Content
Merck
All Photos(1)

Documents

H12005

Sigma-Aldrich

1,6-Hexanedithiol

96%

Synonym(s):

1,6-Dimercaptohexane, DMH, Hexamethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)6SH
CAS Number:
Molecular Weight:
150.31
Beilstein:
1732507
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

~1 mmHg ( 20 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

118-119 °C/15 mmHg (lit.)

mp

−21 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

SCCCCCCS

InChI

1S/C6H14S2/c7-5-3-1-2-4-6-8/h7-8H,1-6H2

InChI key

SRZXCOWFGPICGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,6-Hexanedithiol (HDT) is a long chained bi-functional alkanethiol with 6 carbon atoms structured in a zig-zag pattern, which forms a tetrahedral configuration. It is mainly used as a self-assembled monolayer (SAM) as it functionalizes various surfaces and immobilizes a variety of nanomaterials.

Application

HDT can be used to functionalize molybdenum disulphide (MoS2), which can be used as a two dimensional transition metal dichalcogenide (TMD) in electronics and optoelectronic devices. It forms a SAM on gallium arsenic (GaAs) doped cadmium sulfide (CdS) quantum dots (QDs), which can be used for biosensors and solar applications. It can also be used to immobilize gold nanoparticles (AuNPs) for potential usage in biomedical and bioanalytical applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of biocatalytic enlargement of gold nanoparticles using dynamic light scattering and atomic force microscopy.
Ramanaviciene A, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 510(4), 183-189 (2016)
Thin film fabrication of CdS quantum dots on GaAs substrate by surfactant self-assembly.
Park S and Kongb H
Journal of Ceramic Processing Research, 19(4), 337-341 (2018)
Theoretical and experimental investigation of conjugation of 1, 6-hexanedithiol on MoS2.
Gul A, et al.
Materials Research Express, 5(3), 036415-036415 (2018)
1, 6-hexanedithiol monolayers on Au (111): A multitechnique structural study.
Leung TYB, et al.
Langmuir, 16(2), 549-561 (2000)
A regenerating self-assembled gold nanoparticle-containing electrochemical impedance sensor.
Mahmoud AM, et al.
Biosensors And Bioelectronics, 56(2), 328-333 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service