Skip to Content
Merck
All Photos(1)

Key Documents

01042

Sigma-Aldrich

N-Acetyl-L-cysteine methyl ester

≥90% (HPLC)

Synonym(s):

N-acetyl-L-cysteine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO3S
CAS Number:
Molecular Weight:
177.22
Beilstein:
1724357
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (HPLC)

form

powder or crystals

optical activity

[α]20/D −24.0±3°, c = 1% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

color

white to faint yellow

mp

77-81 °C

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](CS)NC(C)=O

InChI

1S/C6H11NO3S/c1-4(8)7-5(3-11)6(9)10-2/h5,11H,3H2,1-2H3,(H,7,8)/t5-/m0/s1

InChI key

QTKAQJWFVXPIFV-YFKPBYRVSA-N

Other Notes

Sulfur transfer agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renu A Kowluru et al.
Laboratory investigation; a journal of technical methods and pathology, 96(10), 1040-1049 (2016-07-28)
Diabetes elevates matrix metalloproteinase-9 (MMP-9) in the retina and its capillary cells, and activated MMP-9 damages mitochondria, accelerating retinal capillary cell apoptosis, a phenomenon which precedes the development of retinopathy. Diabetes also favors epigenetic modifications regulating the expression of many
A. Gilman et al.
Tetrahedron Letters, 33, 1751-1751 (1993)
Shinji Hatakeyama et al.
Skeletal muscle, 6, 26-26 (2016-07-28)
Cachexia affects the majority of patients with advanced cancer and is associated with reduced treatment tolerance, response to therapy, quality of life, and life expectancy. Cachectic patients with advanced cancer often receive anti-cancer therapies against their specific cancer type as
Jianfen Guo et al.
The Journal of biological chemistry, 286(8), 6500-6509 (2010-12-16)
Protein kinase D (PKD) exists as a family of structurally related enzymes that are activated through similar phosphorylation-dependent mechanisms involving protein kinase C (PKC). While individual PKD isoforms could in theory mediate distinct biological functions, previous studies identify a high
Amy E M Beedle et al.
Nature communications, 9(1), 3155-3155 (2018-08-10)
Mechanical force modifies the free-energy surface of chemical reactions, often enabling thermodynamically unfavoured reaction pathways. Most of our molecular understanding of force-induced reactivity is restricted to the irreversible homolytic scission of covalent bonds and ring-opening in polymer mechanophores. Whether mechanical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service