Skip to Content
Merck
All Photos(3)

Key Documents

A8199

Sigma-Aldrich

N-Acetyl-L-cysteine

≥99% (TLC), suitable for cell culture, BioXtra

Synonym(s):

LNAC, NAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
163.19
Beilstein:
1724426
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)

product line

BioXtra

Quality Level

Assay

≥99% (TLC)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.002% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.5%

color

white to off-white

mp

106-108 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](CS)C(O)=O

InChI

1S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10H,2H2,1H3,(H,6,7)(H,8,9)/t4-/m0/s1

InChI key

PWKSKIMOESPYIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-L-cysteine has been used to study its protective effect against styrene-induced cochlear injuries in rats. It has also been used as a supplement in basal chemically-defined medium (BDM) for the culture of oligodendrocyte precursor cells.

Biochem/physiol Actions

Antioxidant and mucolytic agent. Increases cellular pools of free radical scavengers. Reported to prevent apoptosis in neuronal cells but induce apoptosis in smooth muscle cells. Inhibits HIV replication. May serve as a substrate for microsomal glutathione transferase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective effect of N-acety-L-cysteine (L-NAC) against styrene-induced cochlear injuries
Wei Ping Yang
Acta Oto-Laryngologica, 129(10), 1036-1043 (2009)
Histamine Receptor 3 negatively regulates oligodendrocyte differentiation and remyelination
Chen Y
PLoS ONE, 12(12), e0189380-e0189380 (2017)
Suzanne M Cloonan et al.
Journal of medicinal chemistry, 58(11), 4494-4505 (2015-05-12)
Ruthenium polypyridyl complexes show great promise as new photodynamic therapy (PDT) agents. However, a lack of detailed understanding of their mode of action in cells poses a challenge to their development. We have designed a new Ru(II) PDT candidate that
Michael Berk et al.
Trends in pharmacological sciences, 34(3), 167-177 (2013-02-02)
N-Acetylcysteine (NAC) targets a diverse array of factors germane to the pathophysiology of multiple neuropsychiatric disorders including glutamatergic transmission, the antioxidant glutathione, neurotrophins, apoptosis, mitochondrial function, and inflammatory pathways. This review summarises the areas where the mechanisms of action of
Taeshin Kim et al.
Endocrinology, 156(6), 2185-2199 (2015-04-08)
The appropriate control of synthesis and secretion of the gonadotropin hormones LH and FSH by pituitary gonadotropes is essential for the regulation of reproduction. The hypothalamic neuropeptide GnRH is the central regulator of both processes, coordinating secretion with transcription and

Articles

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

See All

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service