Skip to Content
Merck
All Photos(1)

Documents

FPCR-RO

Roche

FastStart PCR Master

sufficient for ≤100 reactions (04710436001), sufficient for ≤400 reactions (04710444001), sufficient for ≤2,000 reactions (04710452001)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for ≤2,000 reactions (04710452001)
sufficient for ≤100 reactions (04710436001)
sufficient for ≤400 reactions (04710444001)

Quality Level

packaging

pkg of 2 × 1.25 mL (04710436001)
pkg of 8 × 1.25 mL (04710444001)
pkg of 10 × 5 mL (04710452001)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

storage temp.

−20°C

General description

Hot start polymerase chain reaction (PCR) is effortless using the FastStart PCR Master, a ready-to-use, double-concentrated hot start master mix. This master mix contains FastStart Taq DNA Polymerase, Roche high quality PCR-Grade nucleotides and all other reagents required for PCR and two-step reverse transcriptase (RT)-PCR on thermal cycler instruments.
FastStart PCR Master accepts modified nucleotides such as Digoxigenin (DIG)-dUTP. Labeling with DIG-dUTP can be achieved by adding the modified nucleotide in a final concentration of 10 μM to the final volume.
Combine this master mix with Roche′s Transcriptor First Strand cDNA Synthesis Kit to achieve excellent results in two-step RT-PCR.

Application

FastStart PCR Master has been used for the analysis of Nosema sp. DNA by polymerase chain reaction (PCR) method, for validating the genomic DNA by PCR, for amplification of RNA by PCR method.
The FastStart PCR Master is a convenient alternative to the single reagents for nearly all hot start PCR applications. It can be applied for the amplification of genomic DNA and cDNA targets up to 2 kb long, or longer fragments on plasmid DNA targets, with high specificity, sensitivity, and yield. The enzyme is inactive at +15 to +25°C during PCR setup, and then activated at +95°C during initial denaturation. It can be used for:
  • Hot Start PCR up to 2 kb
  • Hot Start RT-PCR up to 2 kb
  • High-throughput PCR
  • Direct colony PCR

Features and Benefits

The FastStart Master has modified recombinant FastStart Taq DNA Polymerase, inactive at temperatures below +75°C. The kit also contains an optimized PCR buffer and GC-RICH Solution for handling a wide range of templates.

  • Higher specificity, sensitivity, and yield:
Hot start PCR makes PCR setup easier.
  • Use robotic setup.
Use this enyzme mix stable for 24 hours at +15 to +25°C.
  • Reduce Setup time.
Store master mixes at +2 to +8°C for immediate use for up to one month.

Contents
2x concentrated master mix containing FastStart Taq DNA Polymerase, Reaction Buffer, and Nucleotides (dATP, dCTP, dGTP, dTTP).

Packaging

1 kit containing: 2x concentrated master mix containing FastStart Taq DNA Polymerase, Reaction Buffer, and Nucleotides (dATP, dCTP, dGTP, dTTP).

Unit Definition

Volume Activity: >= 80U/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

NOTICE TO PURCHASER: LIMITED LICENSE<br />Use of this product is covered by US Patent No. 6,127,155 and corresponding patent claims outside the US.  The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research.  No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel.  This product is for research use only. Human and veterinary diagnostic uses under Roche patent claims require a separate license from Roche.  All uses other than internal research and human and veterinary diagnostic uses under Roche patent claims require a separate license from Thermo Fisher Scientific. Further information on purchasing licenses from Roche may be obtained by contacting the Licensing Department of Roche Molecular Systems, Inc., 4300 Hacienda Drive, Pleasanton, California 94588, USA or Roche Diagnostics GmbH, Sandhofer Strasse 116, 68305 Mannheim, Germany.  Further information on purchasing licenses from Thermo Fisher Scientific may be obtained by contacting the Licensing Department of  Thermo Fisher Scientific, 5791 Van Allen Way, Carlsbad, California 92008, USA.</ br>NOTICE TO PURCHASER: LIMITED LICENSE<br />Use of this product is covered by US Patent Nos. 5,677,152 and 5,773,258, and corresponding patent claims outside the US.  The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research.  No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel.  This product is for research use only.  Human and veterinary diagnostic uses under Roche patent claims require a separate license from Roche.  All uses other than internal research and human and veterinary diagnostic uses under Roche patent claims require a separate license from Thermo Fisher Scientific. Further information on purchasing licenses from Roche may be obtained by contacting the Licensing Department of Roche Molecular Systems, Inc., 4300 Hacienda Drive, Pleasanton, California 94588, USA or Roche Diagnostics GmbH, Sandhofer Strasse 116, 68305 Mannheim, Germany.  Further information on purchasing licenses from Thermo Fisher Scientific may be obtained by contacting the Licensing Department of  Thermo Fisher Scientific, 5791 Van Allen Way, Carlsbad, California 92008, USA.
FastStart is a trademark of Roche

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Kyle Satterstrom et al.
Methods in enzymology, 543, 141-163 (2014-06-14)
Sirtuin 3 (SIRT3) is a major regulator of oncometabolism. Indeed, the activity of SIRT3 significantly affects the response to oxidative stress, glycolytic proficiency, and tumorigenic potential of malignant cells. Thus, a system to accurately measure the transcriptional activity of the
A recurrent point mutation in PRKCA is a hallmark of chordoid gliomas
<BIG>Rosenberg S, et al.</BIG>
Nature Communications, 9, 2371-2371 (2018)
Randy F Stout et al.
PloS one, 8(11), e81771-e81771 (2013-12-07)
Transgene technology is one of the most heavily relied upon tools in modern biological research. Expression of an exogenous gene within cells, for research and therapeutic applications, nearly always includes promoters and other regulatory sequences. We found that repeats of
Laura Jara et al.
Journal of invertebrate pathology, 110(1), 8-13 (2012-02-14)
The recent decline in honey bee colonies observed in both European countries and worldwide is of great interest and concern, although the underlying causes remain poorly understood. In recent years, growing evidence has implicated parasites and pathogens in this decline
First Identification of Nosema Ceranae (Microsporidia) Infecting Apis Mellifera in Venezuela
<BIG>Porrini LP, et al.</BIG>
Journal of Apicultural Science, 61 (2017)

Articles

PCR troubleshooting protocols require exploring all potential error sources to ensure reliable results, addressing each problem independently.

PCR troubleshooting protocols require exploring all potential error sources to ensure reliable results, addressing each problem independently.

PCR troubleshooting protocols require exploring all potential error sources to ensure reliable results, addressing each problem independently.

PCR troubleshooting protocols require exploring all potential error sources to ensure reliable results, addressing each problem independently.

See All

Related Content

Efficiently amplify your samples with confidence using KOD One™ PCR Master Mix for ultra-fast PCR with one of the fastest elongation rates available for high fidelity polymerases.

Efficiently amplify your samples with confidence using KOD One™ PCR Master Mix for ultra-fast PCR with one of the fastest elongation rates available for high fidelity polymerases.

Efficiently amplify your samples with confidence using KOD One™ PCR Master Mix for ultra-fast PCR with one of the fastest elongation rates available for high fidelity polymerases.

Efficiently amplify your samples with confidence using KOD One™ PCR Master Mix for ultra-fast PCR with one of the fastest elongation rates available for high fidelity polymerases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service