Merck
  • Luciferase-based reporter to monitor the transcriptional activity of the SIRT3 promoter.
Luciferase-based reporter to monitor the transcriptional activity of the SIRT3 promoter.
Methods in enzymology (2014-06-14)
F Kyle Satterstrom, Marcia C Haigis
ABSTRACT
Sirtuin 3 (SIRT3) is a major regulator of oncometabolism. Indeed, the activity of SIRT3 significantly affects the response to oxidative stress, glycolytic proficiency, and tumorigenic potential of malignant cells. Thus, a system to accurately measure the transcriptional activity of the SIRT3 promoter could facilitate the identification of novel antineoplastic agents or have diagnostic applications. Here, we describe all the steps involved in the development of a luciferase-based reporter system to measure the activation of the human SIRT3 promoter, encompassing the design of appropriate primers, the cloning of the promoter fragment, and its site-directed mutagenesis. We validated this system in human embryonic kidney 293T cells, taking advantage of the renowned ability of the transcription factor estrogen-related receptor α to transactivate SIRT3. Moreover, here we demonstrate that SIRT3 expression is responsive to rapamycin, a small inhibitor of mammalian target of rapamycin that has been extensively employed as a caloric restriction mimetic. Finally, we provide an overview of the complementary molecular biology techniques that might be employed to further verify the reliability of this system.
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Roche
FastStart PCR Master, sufficient for ≤100 reactions (04710436001), sufficient for ≤400 reactions (04710444001), sufficient for ≤2,000 reactions (04710452001)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution