Skip to Content
Merck
All Photos(2)

Documents

FSTMPMMRO

Roche

FastStart TaqMan® Probe Master

sufficient for 100 reactions, sufficient for 500 reactions, sufficient for 2000 reactions, suitable for qPCR, suitable for RT-qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 100 reactions
sufficient for 2000 reactions
sufficient for 500 reactions

Quality Level

feature

dNTPs included: no
hotstart

manufacturer/tradename

Roche

packaging

pkg of 100 x 50 μL reactions (04673409001)
pkg of 2000 x 50 μL reactions (04673433001)
pkg of 500 x 50 μL reactions (04673417001)

technique(s)

RT-qPCR: suitable
qPCR: suitable

input

purified DNA

detection method

probe-based

General description

FastStart TaqMan® Probe Master is a ready-to-use hot start reaction mix without ROX for quantitative polymerase chain reaction (qPCR) and reverse transcription (RT)-qPCR on real-time PCR systems other than the LightCycle® instruments. It is a 2x concentrated master mix that contains all the reagents (except primers, probe, and template). This master mix simplifies the preparation of reactions for DNA detection and analysis. It allows very sensitive detection and quantification of defined DNA sequences.
Hot start protocols have been also been shown to significantly improve the specificity and yield of PCR. Heat-labile blocking groups on some of the amino acid residues of FastStart Taq DNA Polymerase make the modified enzyme inactive at room temperature. Therefore, there is no elongation during the period when primers can non-specifically bind. The FastStart Taq DNA Polymerase is activated by removing the blocking groups at a high temperature (i.e., a pre-incubation step at +95°C).
This FastStart TaqMan® Probe Master can be used for the amplification and detection of any DNA or cDNA target, including those that are GC- or AT-rich. However, you would need to adapt your detection protocol to the reaction conditions of the particular real-time PCR instrument used and design a specific hydrolysis probe and PCR primers for each target.
Combination of this master mix with our Transcriptor First Strand cDNA Synthesis Kit achieves excellent results in two-step qRT-PCR.
Hot start protocols have been shown to significantly improve the specificity, sensitivity, and yield of PCR. Heat-labile blocking groups on some of the amino acid residues of FastStart Taq DNA Polymerase make the modified enzyme inactive at room temperature. Therefore, there is no elongation during the period when primers can nonspecifically bind. The FastStart Taq DNA Polymerase is activated by removing the blocking groups at a high temperature (i.e., a pre-incubation step at +95°C).

Application

The FastStart TaqMan® Probe Master has been used in:
  • quantitative, real-time DNA-detection assays
  • qPCR and two-step reverse transcription (RT)-qPCR
  • in the hydrolysis probe detection format

Features and Benefits

  • Increase qPCR sensitivity and specificity:
Produce lower cycle threshold (Ct) values with the hot start enzyme FastStart Taq DNA Polymerase and highest purity nucleotides included in the master mix.
  • Use it with any probe-based assay:
Achieve sensitive, specific results in assays with the Universal ProbeLibrary Probes or any other hydrolysis probe.
  • Amplify and detect a broad range of DNA or cDNA targets:
Amplify fragments up to 500 bp long, including those that are GC- or AT-rich.
  • Use with any real-time qPCR instrument other than the LightCycler® Instruments:
Choose from two formulations - one that contains the ROX reference dye and one without ROX.
  • Visualize amplification products on agarose gels.

  • Use robotic pipetting stations to set up qPCR reactions:
Benefit from a master mix that is stable at room temperature during extended reaction setup times.
  • Prevent carryover contamination:
The mix contains dUTP, so that it may be used with Uracil-DNA Glycosylase to prevent false positives arising from carryover contamination (i.e., contamination with amplified DNA).

Components

FastStart TaqMan® Probe Master, 2x concentrated master mix that contains FastStart Taq DNA Polymerase, Reaction Buffer, and Nucleotides (dATP, dCTP, dGTP, dUTP).

Quality

Function test: Each lot is tested for performance in qPCR using three templates: a GC-rich template, an AT-rich template, and a long template (approximately 440 bp).

Other Notes

qPCR targets
In principle, the FastStart TaqMan® Probe Master can be used for the amplification and detection of any DNA or cDNA target, including those that are GC- or AT-rich. However, for each target, you would need:

  • to adapt your detection protocol to the reaction conditions of your real-time PCR instrument, and
  • design specific PCR primers and hydrolysis probes for the target.

See the Operator′s Manual of your real-time PCR instrument for general recommendations.
Two forms available
The master mix is available in two forms – one that contains the ROX reference dye and one without ROX.
Preventing carryover contamination
This master contains dUTP, which will be incorporated into PCR products to help prevent false positives resulting from carryover contamination. In subsequent PCRs, you can add Uracil-DNA Glycosylase to degrade any uracil-containing carryover contaminants (amplification products from previous PCRs).
qRT-PCR
Combine this master mix with our Transcriptor First Strand cDNA Synthesis Kit for two-step qRT-PCR. This kit gives excellent results and works efficiently with all real-time PCR instruments.
For life science research only. Not for use in diagnostic procedures.

Legal Information

LightCycler is a registered trademark of Roche
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ULK1 and ULK2 Regulate Stress Granule Disassembly Through Phosphorylation and Activation of VCP/p97
<BIG>Wang B, et al.</BIG>
Molecular Cell, 74, 742-757 (2019)
Synergistic induction of NY-ESO-1 antigen expression by a novel histone deacetylase inhibitor, valproic acid, with 5-aza-2′-deoxycytidine in glioma cells.
Oi S, et al.
Journal of Neuro-Oncology, 92(1), 15-22 (2009)
Expression of proinflammatory cytokines in osteoarthritis of the temporomandibular joint.
Vernal R, et al.
Archives of Oral Biology, 53(10), 910-915 (2008)
Bo Wang et al.
Molecular cell, 74(4), 742-757 (2019-04-14)
Disturbances in autophagy and stress granule dynamics have been implicated as potential mechanisms underlying inclusion body myopathy (IBM) and related disorders. Yet the roles of core autophagy proteins in IBM and stress granule dynamics remain poorly characterized. Here, we demonstrate

Articles

PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.

PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.

PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.

PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.

See All

Related Content

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service