Skip to Content
Merck
All Photos(1)

Key Documents

10107921001

Roche

Nuclease S7

Micrococcal nuclease, from Staphylococcus aureus

Synonym(s):

s7 nuclease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

Staphylococcus aureus

Quality Level

form

lyophilized

specific activity

~15,000 U/mg

packaging

pkg of 15,000 U

manufacturer/tradename

Roche

optimum pH

7.5

shipped in

wet ice

Application

  • Removal of endogenous RNA from in vitro translation systems[1][2]
  • RNA sequencing experiments
Nuclease S7 has been used in ribonuclease selection process for ribosome profiling.[3][4]

Biochem/physiol Actions

Nuclease S7 is an endonuclease that mainly cleaves single-stranded substrates. It also cleaves double-stranded DNA or RNA.[5] It cleaves the 5′-phosphodiester bonds of RNA and DNA to yield 3′-mononucleotides and oligonucleotides. Activity of the enzyme is dependent on Ca2+ and can be completely inactivated by the addition of EDTA.[6] Nuclease S7 cleavage is highly specific to AT rich region than GC on the DNA.[7]

Unit Definition

One unit releases a sufficient amount of acid-soluble oligonucleotides to produce an increase of 1.0 at A260.

Storage and Stability

Store unopened reagent at 2-8 °C; when diluted to 1 mg/mL, store at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribonuclease selection for ribosome profiling.
Gerashchenko M V and Gladyshev V N
Nucleic Acids Research, 45(2), e6-e6 (2016)
Maxim V Gerashchenko et al.
Nucleic acids research, 49(2), e9-e9 (2020-12-03)
There has been a surge of interest towards targeting protein synthesis to treat diseases and extend lifespan. Despite the progress, few options are available to assess translation in live animals, as their complexity limits the repertoire of experimental tools to
Reimo Zoschke et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(13), E1678-E1687 (2015-03-17)
Chloroplast genomes encode ∼ 37 proteins that integrate into the thylakoid membrane. The mechanisms that target these proteins to the membrane are largely unexplored. We used ribosome profiling to provide a comprehensive, high-resolution map of ribosome positions on chloroplast mRNAs
Ariane Lismer et al.
Nucleic acids research, 48(20), 11380-11393 (2020-10-18)
Advancing the molecular knowledge surrounding fertility and inheritance has become critical given the halving of sperm counts in the last 40 years, and the rise in complex disease which cannot be explained by genetics alone. The connection between both these
High sequence specificity of micrococcal nuclease.
Dingwall C, et al.
Nucleic Acids Research, 9(12), 2659-2674 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service