Skip to Content
Merck
All Photos(1)

Key Documents

04536282001

Roche

DNase I recombinant

grade I, from bovine pancreas, expressed in Pichia pastoris

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.28
form:
lyophilized
solubility:
water: soluble
storage temp.:
2-8°C
Pricing and availability is not currently available.

biological source

bovine pancreas

Quality Level

recombinant

expressed in Pichia pastoris

form

lyophilized

specific activity

(>10,000 U/bottle according to Kunitz (+25°C; DNA as substrate).)

mol wt

39  kDa

packaging

pkg of 2 × 10,000 U

manufacturer/tradename

Roche

concentration

≥70-90%

technique(s)

protein extraction: suitable

color

white to light yellow

solubility

water: soluble

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

application(s)

sample preparation

foreign activity

Proteases, none detected
RNase, none detected

storage temp.

2-8°C

Gene Information

bovine ... DNASE1(282217)

General description

Properties: Double-strand-specific endonuclease that requires bivalent cations for maximal activity.
Activity: 10,000U/bottle according to Kunitz (+25°C; DNA as substrate).
Unit definition: One unit is the enzyme activity that causes an increase in the absorbance of 0.001 per minutes under assay conditions.
Degrades Single- and Double-Stranded DNA; Double Stranded activity requires bivalent cations for maximal activity.

Application

  • DNase I, recombinant, grade I is used for:Eliminating DNA during protein isolation procedures[1]
  • Analysis of chromatin structure
  • Eliminating DNA during sample preparation[2]

Features and Benefits

Contents
Lyophilizate

Unit Definition

One unit, according to Kunitz, is the enzyme activity that under assay conditions causes an absorbance increase at 260 nm of 0.001/minute in 1 ml. Volume activity is determined in 0.1 M sodium acetate, 5 mM MgSO4, pH 5.0.
Assay mixture: Calf thymus DNA (100 μg) is incubated with 20 to 50 U DNase I at +25 °C.
The increase in absorbance is measured at 260 nm.

Information Note: These conditions are used for the determination of activity,according to Kunitz, since the Kunitz assay results in high reproducibility and sensitivity. When using this enzyme for normal experimental purposes, Roche recommends using incubation buffers that are appropriate for a given application, e.g., as mentioned in the standard literature (Sambrook et al., Curr. Prot. Mol. Biol., etc.).

Preparation Note

Activator: The enzyme requires divalent cations for maximal activity. The specificity of the reaction depends on the nature of the cation used. In the presence of Mg2+ DNase I causes single-stranded nicks in dsDNA, while in the presence of Mn2+ the enzyme produces double-stranded breaks.
Storage conditions (working solution): Long-term Storage of the Dissolved Enzyme

The solvent generally recommended for DNase I is water. When DNase I is reconstituted in water, the solution can be kept for 2 days at 2 to 8 °C and for 1 month at -15 to -20 °C. For best results, prepare appropriate aliquots and avoid repeated freezing and thawing. However, for long-term storage, carefully dissolve DNase I in one of the buffers listed below. In buffer 1, the enzyme will be stable for several weeks. The enzyme will be stable for at least 18 months if it is dissolved in buffer 2, 3, or 4 and stored at the recommended temperatures.

Information note: Do not vortex the enzyme while dissolving!
Buffer 1: 20 mM Tris-HCl, 20 mM MgCl2, 5 mM CaCl2, 0.1 mM dithioerythritol, 0.1 mM EDTA, 50% (v/v) glycerol, pH 8. Store this solution at -15 to-25 °C; it will not freeze at this temperature and will be stable for several weeks.
Buffer 2: 20 mM Tris-HCl, 50 mM NaCl, 1 mM dithioerythritol, 100 μg/mL BSA, 50% glycerol (v/v), pH 7.6. Store this solution at -15 to -25 °C for up to 18 months; it will not freeze at this temperature.
Buffer 3: 20 mM Tris-HCl, 1 mM MgCl2, 50% (w/v) glycerol, pH 7.5. Store this solution at -15 to -25 °C for up to 18 months; it will not freeze at this temperature.
Buffer 4: 20 mM Tris-HCl, 1 mM MgCl2, pH 7.5. Aliquot in appropriate amounts (e.g., 10 μl), freeze the aliquots quickly on dry ice, and store at -60 °C or below for up to 18 months. Thaw only the amount needed for each experiment. Do not refreeze. Discard any leftover thawed solution.

Storage and Stability

Tightly closed. Dry.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delia Alba Soto et al.
Reproduction (Cambridge, England), 161(3), 239-253 (2020-12-05)
The germ cell lineage ensures the creation of new individuals and perpetuates the genetic information across generations. Primordial germ cells are pioneers of gametes and exist transiently during development until they differentiate into oogonia in females, or spermatogonia in males.
Xuedan He et al.
Journal for immunotherapy of cancer, 9(12) (2021-12-05)
Induction of CD8+ T cells that recognize immunogenic, mutated protein fragments in the context of major histocompatibility class I (MHC-I) is a pressing challenge for cancer vaccine development. Using the commonly used murine renal adenocarcinoma RENCA cancer model, MHC-I restricted
Tomasz Ahrends et al.
STAR protocols, 3(1), 101157-101157 (2022-02-12)
The myenteric plexus is located between the longitudinal and circular layers of muscularis externa in the gastrointestinal tract. It contains a large network of enteric neurons that form the enteric nervous system (ENS) and control intestinal functions, such as motility
Xuedan He et al.
Small (Weinheim an der Bergstrasse, Germany), 17(11), e2007165-e2007165 (2021-02-20)
Human papilloma virus (HPV)-16 is associated with cervical cancers and induces expression of the E6 and E7 oncogenes. Using a murine cell line that expresses these, the genes are sequenced, and six predicted major histocompatibility complex (MHC) class I (MHC-I)
Xuedan He et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(24), e2103023-e2103023 (2021-10-31)
Short peptides reflecting major histocompatibility complex (MHC) class I (MHC-I) epitopes frequently lack sufficient immunogenicity to induce robust antigen (Ag)-specific CD8+ T cell responses. In the current work, it is demonstrated that position-scanning peptide libraries themselves can serve as improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service