Skip to Content
Merck

COLLP-RO

Roche

Collagenase P

from Clostridium histolyticum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Biological source:
Clostridium histolyticum
Concentration:
0.5-2.5 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Clostridium histolyticum

sterility

non-sterile

form

lyophilized

collagenase activity

>1.5 U/mg (Lyophilizate, Collagenase activity)

packaging

pkg of 1 g (11249002001), pkg of 100 mg (11213857001), pkg of 2.5 g (11213873001), pkg of 500 mg (11213865001)

manufacturer/tradename

Roche

concentration

0.5-2.5 mg/mL

technique(s)

tissue processing: suitable

color

light brown to brown

solubility

water: soluble

NCBI accession no.

application(s)

life science and biopharma

foreign activity

Clostripain 24.099 U/mg, Protease 4.4 U/mg (Proteases), Trypsin 53.428 U/mg (with BAEE)

storage temp.

2-8°C

General description

Collagenase P is an enzyme mixture prepared from the extracellular culture filtrate of a special Clostridium histolyticum strain.Collagenase P is a research biochemical, not manufactured under the US good manufacturing practices (GMP) regulations.
Note: Collagenase P is a non-sterile preparation of the culture supernatant of Clostridium histolyticum. It is neither designed nor intended for isolation of pancreatic islets for transplantation into humans and must not be used for such purposes.

Application

Collagenase from C. histolyticum is used for the dissociation of tissues for the establishment of primary cell cultures. Because of its high collagenase activity Collagenase P is particularly suitable for the isolation of pancreatic islets from mouse and rat (function tested). The preparation is also suitable for the isolation of adipocytes from epididymal fat pads of rats.

Biochem/physiol Actions

Enzyme activity:
Collagenase activity: >1.5U/mg (according to Wünsch) (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate)
Contaminating enzyme activities: trypsin, clostripain, and total proteolytic activity
Collagenase P has a very high collagenase activity (>1.5U/mg), and is function tested for the isolation of mouse pancreatic islets.
Specificity: Collagenase degrades native collagen. Clostripain, trypsin-like enzymes, and neutral proteases degrade other proteins as well.

Features and Benefits

Contents:
Lyophilizate, nonsterile

Preparation Note

Activator: Ca2+
Inhibitors:
Collagenase inhibitors: EDTA, EGTA, Cys, His, DTT, 2-mercaptoethanol
Collagenase is not inhibited by serum.
Clostripain inhibitors: TLCK
Trypsin inhibitors: aprotinin, trypsin inhibitor (egg white, soybean)
Working concentration: 0.5 to 2.5mg/ml
0.5 to 1.5mg/ml for the isolation of pancreatic islets from mouse and rat pancreas.
Approximately 2mg/ml for the isolation of adipocytes.
Storage conditions (working solution): -15 to -25°C
The reconstituted solution is stable at -15 to -25°C. Prepare appropriate aliquots and avoid repeated freezing and thawing.
Reconstitution in any balanced salt solution (e.g., HBSS)
Store dry

Other Notes

For life science research only. Not for use in diagnostic procedures.


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



T-cadherin (Cdh13) in association with pancreatic ?-cell granules contributes to second phase insulin secretion.
Tyrberg B
Islets, 3(6), 327-337 (2011)
Luigi Racioppi et al.
The Journal of biological chemistry, 287(14), 11579-11591 (2012-02-16)
Calcium/calmodulin-dependent kinase kinase 2 (CaMKK2) plays a key role in regulating food intake and energy expenditure at least in part by its actions in hypothalamic neurons. Previously, we showed that loss of CaMKK2 protected mice from high-fat diet (HFD)-induced obesity
Fumin Lin et al.
Endocrinology, 152(10), 3668-3679 (2011-08-25)
When fed a standard chow diet, CaMKK2 null mice have increased adiposity and larger adipocytes than do wild-type mice, whereas energy balance is unchanged. Here, we show that Ca(2+)/calmodulin-dependent protein kinase kinase 2 (CaMKK2) is expressed in preadipocytes, where it



Global Trade Item Number

SKUGTIN
1121385700104061838699695
1121387300104061838250742
1121386500104061838250735
1124900200104061835508419