Skip to Content
Merck

SCM011

Sigma-Aldrich

ENStem-A Neural Freezing Medium (1X)

The ENStem-A Neural Freezing Medium is qualified for use with ENStem-A Human Neural Progenitor Cells.

Synonym(s):

Freezing Medium

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$448.92

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

To order products, please contact your local dealer.

Select a Size

Change View
100 μG
$448.92

About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

To order products, please contact your local dealer.

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5200988SAB5200888SAB5200904
conjugate

unconjugated

conjugate

Allophycocyanin conjugate

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse, rat

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

clone

S327A-38, monoclonal

clone

S327A-38, monoclonal

clone

S327-95, monoclonal

clone

S327-95, monoclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

General description

ENStem-A Neural Freezing Medium is qualified for use with ENStem-A Human Neural Progenitor Cells (Part No. SCC003) cultured in defined serum-free conditions with ENStem-A Neural Expansion Medium (Cat. No. SCM004). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.

Storage and Stability

Store at -20°C. Refer to lot expiration date on label.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service