Skip to Content
Merck
All Photos(1)

Key Documents

MAB8188

Sigma-Aldrich

Anti-EBV EA-R-p17 Antibody, clone 5B11

clone 5B11, Chemicon®, from mouse

Synonym(s):

Early Antigen Protein R, Nuclear Antigen, BHRF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
5B11, monoclonal
application:
ELISA
IF
IP
WB
species reactivity:
human
technique(s):
ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
11

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

shipped in

wet ice

General description

The Early Restricted protein (Ea-R) of Epstein-Bar Virus is encoded by the viral BHFR1 gene. Ea-R is a 17 kDa protein (p17) with protein kinase activity. Ea-R-p17 is a functional homologue of the anti-apoptotic oncoprotein bcl-2, which is expressed during the switch from latency to lytic replication and may be involved in proliferation of EBV infected cells. However, Ea-R-p17 does not appear to be essential for virus replication or transformation of B cells in vitro.

Specificity

Recognizes the 17 kDa Early Restricted Protein (Ea-R-p17) of Epstein-Barr Virus by immunoblot, immunoprecipitation and immunofluorescence.

Application

Anti-EBV EA-R-p17 Antibody, clone 5B11 is an antibody against EBV EA-R-p17 for use in ELISA, IF, IP & WB.
Immunoblotting

Immunoprecipitation

Immunofluorescence

Optimal dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin presented as a liquid in 0.02M PBS, 0.25M NaCl, with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative analysis of the expression of the Epstein-Barr virus (EBV) anti-apoptotic gene BHRF1 in nasopharyngeal carcinoma and EBV-related lymphoid diseases
Nicholls, J. et al.
Journal of Medical Virology, 65, 105-113 (2001)
Mingxia Shi et al.
Virus research, 177(1), 11-21 (2013-07-31)
Transition of Akata Burkitt's lymphoma (BL) from a malignant to nonmalignant phenotype upon loss of Epstein-Barr virus (EBV) is evidence for a viral contribution to tumorigenesis despite the tight restriction of EBV gene expression in BL. Examination of global cellular
Functional differences between BHRF1, the Epstein-Barr virus-encoded Bcl-2 homologue, and Bcl-2 in human epithelial cells
Dawson, C. et al.
Journal of virology, 72, 9016-9024 (1998)
C A Meseda et al.
The Journal of general virology, 81(Pt 7), 1801-1805 (2000-06-22)
The human tumour virus Epstein-Barr virus (EBV) encodes a 17 kDa protein, BHRF1, which is a member of the BCL:-2 family and has been shown to suppress apoptosis. The role of this gene in the life-cycle of EBV has not
Epstein-Barr virus contributes to the malignant phenotype and to apoptosis resistance in Burkitt's lymphoma cell line Akata.
J Komano, M Sugiura, K Takada
Journal of virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service