Skip to Content
Merck
All Photos(2)

Key Documents

T3705

Sigma-Aldrich

Transferrin human

Optiferrin, recombinant, expressed in rice

Synonym(s):

Optiferrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

recombinant

expressed in rice

Assay

≥95% (agarose gel electrophoresis)

form

powder

composition

Protein, ≥90%

Iron content

≥300 μg/g

technique(s)

immunoprecipitation (IP): suitable
tissue culture: suitable

impurities

≤1.0 EU/mg Endotoxin

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TF(7018)

General description

Transferrin is an 80 kDa serum glycoprotein, which is expressed in hepatocytes. The gene is located on human chromosome 3q22.1.
Optiferrin is recombinant human transferrin that is completelyblood-free. Optiferrin’s performance is equivalent to human holo transferrin incell culture and offers significant regulatory advantages due to its safety andconsistency. Optiferrin is suitable for a broad range of cell types thatinclude stem, primary, and bioproduction cells.

Application

Transferrin Human has been used to perform immunoprecipitation measurements. It has also been used for the preparation of essential 8 medium.

Biochem/physiol Actions

Human transferrin was shown to promote allogeneic chimerism in mice after irradiation and transplantation of incompatible bone marrow.
Transferrin facilitates the transfer of iron to erythron and peripheral tissues. It stimulates the proliferation of human lymphocytes. Mutations in this gene are linked to atransferrinemia.

Other Notes

1). Patent Rights
U.S. Patent No. 9,321,828
2). Registered Trademark Rights
U.S. Reg. 3,880,881
International Reg. 1,466,509

Legal Information

Optiferrin is a trademark of InVitria

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Axelman et al.
Thrombosis and haemostasis, 112(3), 466-477 (2014-07-18)
Heparanase is implicated in cell invasion, tumour metastasis and angiogenesis. It forms a complex and enhances the activity of the blood coagulation initiator - tissue factor (TF). We describe new peptides derived from the solvent accessible surface of TF pathway
A remote-controlled immunochemical system for nephelometric detection of human serum transferrin
Strzelak K and Koncki R
Biosensors And Bioelectronics, 127(2), 31-37 (2019)
Chemically defined culture and cardiomyocyte differentiation of human pluripotent stem cells
Burridge PW, et al.
Current Protocols in Human Genetics / Editorial Board, Jonathan L. Haines ... [Et al.], 87(1), 21-23 (2015)
TFRC (transferrin receptor (p90, CD71))
Dorak MT
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2009)
Effect of transferrin, lactoferrin and chelated iron on human T-lymphocytes
Djeha A and Brock JH
British Journal of Haematology, 80(2), 235-241 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service