Skip to Content
Merck
All Photos(3)

Key Documents

MAB5502

Sigma-Aldrich

Anti-Vesicular Glutamate Transporter 1 Antibody

CHEMICON®, mouse monoclonal, 3C10.2

Synonym(s):

Solute carrier family 17 member 7, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, vGluT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Vesicular Glutamate Transporter 1 Antibody, clone 3C10.2, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C10.2, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

VGLUT1 is expressed in a subset of glutamate neurons and transports glutamate into native synaptic vesicles from the brain, exhibiting a conductance for chloride that is blocked by glutamate. Vesicular glutamate transport has a substantially lower apparent affinity than the plasma membrane excitatory amino acid transporters. Glutamate transport by VGLUT1 is saturated with a K(m) of approximately 2 mM, in the same range as transport by synaptic vesicles. Finally, plasma membrane glutamate transporters recognize both aspartate and glutamate as substrates, whereas VGLUT1 does not recognize aspartate. Vesicular glutamate transporters pack the neurotransmitter into synaptic vesicles so that they can be released into the synapse. VGLUTs are dependent on a proton gradient that they create by hydrolysing adenosine triphosphate (ATP). VGLUTs have only between one hundredth and one thousandth the affinity for glutamate that EAATs have.

Specificity

Reacts with VGLUT1 (Vesicular Glutamate Transporter 1).

Immunogen

Recombinant protein from rat VGLUT1.

Application

Immunohistochemistry:
A previous lot of this antibody was tested on IH.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuronal & Glial Markers
This Anti-Vesicular Glutamate Transporter 1 Antibody, clone 3C10.2, Cat. No. MAB5502, is validated for use in IH, WB for the detection of VGluT1.

Quality

Routinely evaluated by Western Blot on Mouse Brain lysates.

Western Blot Analysis:
1:500 dilution of this lot detected Vesicular Glutamate Transporter 1 on 10 μg of Mouse Brain lysates.

Target description

60 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 in buffer containing PBS containing 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8ºC in undiluted aliquots from date of receipt.

Analysis Note

Control
Brain

VGLUT1 Control Peptide AG208

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

April P Neal et al.
Toxicological sciences : an official journal of the Society of Toxicology, 116(1), 249-263 (2010-04-09)
Lead (Pb(2+)) exposure is known to affect presynaptic neurotransmitter release in both in vivo and cell culture models. However, the precise mechanism by which Pb(2+) impairs neurotransmitter release remains unknown. In the current study, we show that Pb(2+) exposure during
Kajari Karmakar et al.
Cell reports, 18(1), 185-197 (2017-01-05)
Tonotopy is a hallmark of auditory pathways and provides the basis for sound discrimination. Little is known about the involvement of transcription factors in brainstem cochlear neurons orchestrating the tonotopic precision of pre-synaptic input. We found that in the absence
Olga C Jorge-Torres et al.
Cell reports, 23(6), 1665-1677 (2018-05-10)
Rett syndrome (RTT) is the second leading cause of mental impairment in girls and is currently untreatable. RTT is caused, in more than 95% of cases, by loss-of-function mutations in the methyl CpG-binding protein 2 gene (MeCP2). We propose here
Xiaoyang Li et al.
Stem cells and development, 25(10), 803-813 (2016-03-30)
Spiral ganglion neurons (SGNs) are usually damaged in sensorineural hearing loss. SGN-derived neural stem cells (NSCs) have been identified and proposed to differentiate into neurons to replace damaged SGNs. However, it remains obscure whether SGN-NSC-derived neurons (ScNs) are electrophysiologically functional
Yuanyuan Li et al.
The Journal of biological chemistry, 292(48), 19590-19604 (2017-10-04)
Proper neural commitment is essential for ensuring the appropriate development of the human brain and for preventing neurodevelopmental diseases such as autism spectrum disorders, schizophrenia, and intellectual disorders. However, the molecular mechanisms underlying the neural commitment in humans remain elusive.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service