Skip to Content
Merck
All Photos(1)

Key Documents

MAB4708

Sigma-Aldrich

Anti-Caspase 8 Antibody, clone 1-1-37

clone 1-1-37, Chemicon®, from mouse

Synonym(s):

MACH1, FLICE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1-1-37, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

Not specified

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP8(841)

General description

Caspase-8 is a 55 kDa protein that binds to the death effector domain (DED) of FADD through an analogous DED domain present in the proform of caspase-8. Recruitment of caspase-8 to the Fas receptor results in oligomerization of the caspase-8 protein, which drives its auto-activation through self-cleavage. Active caspase-8 then proteolytically activates other downstream caspases including caspase-9, which lead to cell death.

Specificity

Highly specific to caspase-8 and shows no cross-reaction with other caspase family members. This antibody recognizes the full length 55kDa caspase 8 on western blots.

Immunogen

Recombinant full-length human caspase-8 protein

Application

Detect Caspase 8 using this Anti-Caspase 8 Antibody, clone 1-1-37 validated for use in WB.
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: 1:100 - 1:1000

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-573; 04-574

Physical form

Format: Purified
Purified immunoglobulin by protein A. Liquid in PBS, pH 7.6, with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha Lappas et al.
Molecular human reproduction, 24(2), 94-109 (2017-12-23)
What is the association between placental formyl peptide receptor 2 (FPR2) and trophoblast and endothelial functions in pregnancies affected by foetal growth restriction (FGR)? Reduced FPR2 placental expression in idiopathic FGR results in significantly altered trophoblast differentiation and endothelial function
Jessica J Huck et al.
Molecular cancer research : MCR, 8(3), 373-384 (2010-03-04)
Aurora A kinase is a serine/threonine protein kinase responsible for regulating several mitotic processes including centrosome separation, spindle assembly, and chromosome segregation. Small molecule inhibitors of Aurora A kinase are being pursued as novel anticancer agents, some of which have
Activated Cdc42-associated kinase 1 (Ack1) is required for tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) receptor recruitment to lipid rafts and induction of cell death.
Linderoth, E; Pilia, G; Mahajan, NP; Ferby, I
The Journal of Biological Chemistry null
Padma Murthi et al.
Cells, 9(4) (2020-04-16)
We reported earlier that an anti-inflammatory small peptide receptor-formyl peptide receptor-2 (FPR2) was significantly decreased in placentas from third trimester pregnancies complicated with fetal growth restriction (FGR), compared to placentas from uncomplicated control pregnancies, suggesting FPR2 may play a role
Cheng-Wen Lin et al.
Biomedicines, 8(9) (2020-08-23)
In this study, the anti-tumor activity of ilimaquinone (IQ), a sesquiterpene quinone isolated from marine sponge Halichondria sp., in oral squamous cell carcinoma (OSCC) cells, was investigated. IQ suppressed the viability of the OSCC cell lines SCC4 and SCC2095 with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service