Skip to Content
Merck
All Photos(1)

Key Documents

M0317

Sigma-Aldrich

Microsomes from Liver, Pooled

from human

Synonym(s):

Liver Microsomes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

human

Quality Level

packaging

vial of ~10 mg

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Application

Microsomes from liver have been used to study the in vitro effect of Clinacanthus nutans leaves on the activity of cytochrome P450 3A4 and 2E1.

Biochem/physiol Actions

Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.
Source of drug metabolizing enzymes, including cytochrome P-450.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verawan Uchaipichat
Drug metabolism and pharmacokinetics, 33(1), 67-76 (2017-12-16)
This study aimed to investigate the liver microsomal inhibitory effects of silybin, silychristin, andrographolide, and curcumin by using morphine as an in vitro UGT2B7 probe substrate, and predict the magnitude of the herb-drug interaction arising from these herbal constituents' inhibition in vivo.
Cytotoxicity and cytochrome P450 inhibitory activities of Clinacanthus nutans
Sukh Yen Khua
Drug metabolism and personalized therapy (2017)
Britta Heinze et al.
European journal of nuclear medicine and molecular imaging, 49(1), 301-310 (2021-07-04)
Adrenal tumors represent a diagnostic and therapeutic challenge. Promising results have been obtained through targeting the cytochrome P450 enzymes CYP11B1 and CYP11B2 for molecular imaging, and [123/131I]iodometomidate ([123/131I]IMTO) has even been successfully introduced as a theranostic agent. As this radiopharmaceutical
Anand Joshi et al.
Cancer chemotherapy and pharmacology, 86(4), 535-545 (2020-09-20)
Carboplatin dose is calculated based on kidney function, commonly estimated with imperfect creatinine-based formulae. Iohexol is used to measure glomerular filtration rate (GFR) and allows calculation of a more appropriate carboplatin dose. To address potential concerns that iohexol administered during
Jeffry Adiwidjaja et al.
British journal of clinical pharmacology, 86(10), 2080-2094 (2020-04-07)
This study aimed to investigate the potential interaction between Schisandra sphenanthera, imatinib and bosutinib combining in vitro and in silico methods. In vitro metabolism of imatinib and bosutinib using recombinant enzymes and human liver microsomes were investigated in the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service