Skip to Content
Merck
All Photos(3)

Key Documents

06-520

Sigma-Aldrich

Anti-Na+/K+ ATPase α-1 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-CMT2DD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, rat, human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Na+/K+ ATPase α-1 is a human gene and functions to maintain Na+ and K+ across the cell membrane which is important for osmoregulation. It is a component of (Na+/K+) ATPase, a transporter that catalyzes the hydrolysis of ATP and transports sodium and potassium ions across the plasma membrane. It also partners with Src and other signaling proteins. Binding of ouabain and other cardiotonic steroids to the (Na+/K+) ATPase activates Src, resulting in the assembly and activation of multiple signaling cascades. Binding of ouabain also induces a positive inotropic effect. Diseases associated with ATPase are Thyrotoxic periodic paralysis and hyperthyroidism. (References: Wikipedia and http://www.bioportfolio.com/gene/476-ATP1A1.html)

Specificity

Expected to cross-react with Rat-100%, Mouse-99%; human, pig, rabbit, monkey-96%.
Recognizes the α-1 subunit isoform of Na+/K+ ATPase.

Immunogen

Epitope: Cytoplasmic domain
GST-tagged ATPase.

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Na+/K+ ATPase α-1 Antibody is validated for use in IC, IP, WB for the detection of Na+/K+ ATPase α-1.

Quality

Routinely evaluated by Immunoprecipitation on rat brain microsomal prep. lysate.

Target description

112 kDa

Physical form

Format: Purified
Protein A purified
Purified antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl and 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Rat Brain Microsomal Prep. lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Chung Youn et al.
Scientific reports, 5, 18420-18420 (2015-12-18)
Excessive exposure to club drug (GHB) would cause cognitive dysfunction in which impaired hippocampal Ca(2+)-mediated neuroplasticity may correlate with this deficiency. However, the potential changes of in vivo Ca(2+) together with molecular machinery engaged in GHB-induced cognitive dysfunction has never
Purification of active Na+-K+-ATPase using a new ouabain-affinity column
Yingst, D. R., et al
The American Journal of Physiology, 275, C1167-C1177 (1998)
Antisera specific for the alpha 1, alpha 2, alpha 3, and beta subunits of the Na,K-ATPase: differential expression of alpha and beta subunits in rat tissue membranes.
Shyjan, A W and Levenson, R
Biochemistry, 28, 4531-4535 (1989)
Harindarpal S Gill et al.
Biochimica et biophysica acta, 1850(6), 1286-1296 (2015-03-07)
Band keratopathy, an ocular disease that is characterized by hypercalcemia and opaque bands across the cornea, has been associated with kidney disease. Type-II renal tubular acidosis (RTA), a condition in which the kidneys fail to recover bicarbonate (HCO3-) in the
Insulin-induced recruitment of glucose transporter 4 (GLUT4) and GLUT1 in isolated rat cardiac myocytes. Evidence of the existence of different intracellular GLUT4 vesicle populations
Fischer, Y., et al
The Journal of Biological Chemistry, 272, 7085-7092 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service