Skip to Content
Merck
All Photos(1)

Documents

GF142

Sigma-Aldrich

Platelet Derived Growth Factor-AA Protein, Recombinant human

The Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle & connective tissue.

Synonym(s):

PDGF-AA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

biological source

human

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

technique(s)

cell culture | mammalian: suitable

input

sample type neural stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PDGFA(5154)

General description

Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF, which is composed of a dimer of two chains termed the A chain and B chain, can be present as AA or BB homodimers or as an AB heterodimer. Human PDGF-AA is a 28.5 kDa A chain homodimer protein containing 250 amino acid residues.
Product Source: E.coli

Application

For most in vitro applications, PDGF-AA exerts its biological activity in the concentration range of 0.2 to 10.0 ng/mL. Responding cells are (partial list): Fibroblasts, connective tissue.

Linkage

Replaces: GF017

Physical form

Lyophilized from a 10mM Acetic Acid solution.

Storage and Stability

The lyophilized PDGF-AA is stable for a few weeks at room temperature, but best stored at -80°C. After a quick spin, reconstitute in 10mM Acetic Acid to a concentration of 0.1-1.0 mg/mL. This solution can be diluted into aqueous buffered solutions and stored at 2-8°C for one week or -20°C for future use. Reconstituted PDGF-AA should be stored in undiluted aliquots at -20°C for up to six months.

Analysis Note

Specific Activity: Human PDGF-AA is fully biologically active when compared to standards, as determined by colorimetric assay using BALB / 3T3 cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service