Skip to Content
Merck
All Photos(2)

Key Documents

ABN1362

Sigma-Aldrich

Anti-RBPMS Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

RNA-binding protein with multiple splicing, RBPMS, RBP-MS, Heart and RRM expressed sequence, Hermes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
polyclonal
application:
IF
WB
species reactivity:
human, mouse, rat
technique(s):
immunofluorescence: suitable
western blot: suitable
citations:
28

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBPMS(11030)

General description

RNA-binding protein with multiple splicing (UniProt Q93062; also known as Heart and RRM expressed sequence, Hermes, RBP-MS) is encoded by the RBPMS (also known as HERMES) gene (Gene ID 11030) in human. RBPMS is a member of the RRM (RNA recognition motif) family of RNA-binding proteins. Transcriptome-wide study using photoactivatable-ribonucleoside-enhanced crosslinking and immunoprecipitation (PAR-CLIP) in HEK293 cells reveals both exonic mature and intronic pre-mRNA-binding sites, in agreement with the nuclear and cytoplasmic localization of the RRM family proteins. RBPMS is found to be exclusively expressed in retinal ganglion cells (RGCs) and displaced RGCs (dRGCs). RBPMS expression can therefore be utitized as a RGC marker, and the RBPMS gene promoter could be utilized for driving RGC-specific transgene expression.

Specificity

Expected to react with all five spliced isoforms (UniProt Q93062-1 through Q93062-5).

Immunogen

Epitope: N-terminal region.
KLH-conjugated linear peptide corresponding to a sequence from the N-terminal region of human RBPMS.

Application

Anti-RBPMS Antibody is an antibody against RBPMS for use in Western Blotting, Immunofluorescence.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS
Western Blotting Analysis: A representative lot detected endogenous RBPMS in mouse and rat retina extracts, as well as exogenously expressed human RBPMS in transfected HEK293T cells (Rodriguez, A.R., et al. (2014). J. Comp. Neurol. 522(6):1411-1443).
Immunofluorescence Analysis: A representative lot detected RBPMS immunoreactivity primarily associated with cell bodies located in the ganglion cell layer (GCL) of paraformaldehyde-fixed mouse retinas by fluorescent immunohistochemistry using whole-mounted retinas (Rodriguez, A.R., et al. (2014). J. Comp. Neurol. 522(6):1411-1443).
Immunofluorescence Analysis: An 1:5,000 dilution of a representative lot detected RBPMS immunoreactivity among cells in the ganglion cell layer (GCL) of paraformaldehyde-fixed mouse retinas by fluorescent immunohistochemistry (Courtesy of Dr. Nicholas Brecha, David Geffen School of Medicine at UCLA).

Quality

Evaluated by Western Blotting in mouse embryonic stem cell (mESC) lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RBPMS in 10 µg of mouse embryonic stem cell (mESC) lysate.

Target description

~25 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in PBS with 1% BSA and 0.09 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelsea Guymer et al.
Translational vision science & technology, 9(6), 28-28 (2020-08-22)
The ability to accurately quantify immunohistochemically labeled retinal ganglion cells (RGCs) on wholemounts is an important histopathological determinant in experimental retinal research. Traditionally, this has been performed by manual or semi-automated counting of RGCs. Here, we describe an automated software
Alexandra Bernardo-Colón et al.
Frontiers in neuroscience, 13, 719-719 (2019-07-30)
Indirect traumatic optic neuropathy (ITON) is a condition that is often associated with traumatic brain injury and can result in significant vision loss due to degeneration of retinal ganglion cell (RGC) axons at the time of injury or within the
Chelsea Guymer et al.
Translational vision science & technology, 9(6), 28-28 (2020-08-22)
The ability to accurately quantify immunohistochemically labeled retinal ganglion cells (RGCs) on wholemounts is an important histopathological determinant in experimental retinal research. Traditionally, this has been performed by manual or semi-automated counting of RGCs. Here, we describe an automated software
Chelsea Guymer et al.
Translational vision science & technology, 9(6), 28-28 (2020-08-22)
The ability to accurately quantify immunohistochemically labeled retinal ganglion cells (RGCs) on wholemounts is an important histopathological determinant in experimental retinal research. Traditionally, this has been performed by manual or semi-automated counting of RGCs. Here, we describe an automated software
Lucia G Le Roux et al.
Cells, 9(5) (2020-05-28)
We illuminate a possible explanatory pathophysiologic mechanism for retinal cellular neuropathy by means of a novel diagnostic method using ophthalmoscopic imaging and a molecular imaging agent targeted to fast axonal transport. The retinal neuropathies are a group of diseases with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service