Skip to Content
Merck
All Photos(1)

Documents

SAB2500895

Sigma-Aldrich

Anti-RPE65 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-LCA2, Anti-RP20, Anti-Rd12, Anti-Retinal pigment epithelium-specific protein 65 kDa, Anti-SRPE65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPE65(6121)

Immunogen

Peptide with sequence C-EVKKNARKAPQPE from the internal region of the protein sequence according to NP_000320.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianing Gu et al.
Acta biomaterialia, 92, 115-131 (2019-05-11)
In vitro generation of a functional retinal pigment epithelium (RPE) monolayer sheet is useful and promising for RPE cell therapy. Here, for the first time, we used induced pluripotent stem (iPS) supernatant as the conditioned medium (iPS-CM) and femtosecond laser
Xiaokun Wang et al.
Journal of tissue engineering and regenerative medicine, 12(3), 821-829 (2017-10-20)
Structural and biochemical cues of extracellular matrix can substantially influence the differentiation and maturation of cultured retinal pigment epithelial (RPE) cells. In this study, thin collagen vitrigels were engineered to create collagen nanofibrillar structures of different fibril densities in an
Yan Shao et al.
American journal of physiology. Endocrinology and metabolism, 316(3), E443-E452 (2018-12-24)
The purpose of this study was to explore the role of microRNA-451a (miR-451a) in diabetic retinopathy through activating transcription factor 2 (ATF2). The epiretinal membrane samples from patients with proliferative diabetic retinopathy (PDR) were immunolabeled with an antibody for Ki-67
Yan Li et al.
Human mutation, 40(4), 426-443 (2019-01-11)
Human RPE65 mutations cause a spectrum of retinal dystrophies that result in blindness. While RPE65 mutations have been almost invariably recessively inherited, a c.1430A>G (p.(D477G)) mutation has been reported to cause autosomal dominant retinitis pigmentosa (adRP). To study the pathogenesis
Yuqiong Hu et al.
PLoS biology, 17(7), e3000365-e3000365 (2019-07-04)
The developmental pathway of the neural retina (NR) and retinal pigment epithelium (RPE) has been revealed by extensive research in mice. However, the molecular mechanisms underlying the development of the human NR and RPE, as well as the interactions between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service