8.41268
Trimethoxyvinylsilane
for synthesis
Synonym(s):
Trimethoxyvinylsilane, Vinyltrimethoxysilane
About This Item
Recommended Products
vapor pressure
88 hPa ( 55 °C)
Quality Level
Assay
≥97.0% (GC)
form
liquid
autoignition temp.
235 °C
potency
7120 mg/kg LD50, oral (Rat)
3259 mg/kg LD50, skin (Rabbit)
expl. lim.
1.1 % (v/v)
bp
123 °C/1013 hPa
transition temp
flash point 22 °C
density
0.97 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[Si](OC)(OC)(OC)C=C
InChI
1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3
InChI key
NKSJNEHGWDZZQF-UHFFFAOYSA-N
Related Categories
Analysis Note
Density (d 20 °C/ 4 °C): 0.968 - 0.973
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Flam. Liq. 2 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
71.6 °F - closed cup
Flash Point(C)
22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service