Skip to Content
Merck
All Photos(1)

Key Documents

540411

Sigma-Aldrich

Puromycin, Dihydrochloride, Cell Culture-Tested

Puromycin, CAS 58-58-2, is a protein synthesis inhibitor that causes premature release of nascent polypeptide chains.

Synonym(s):

Puromycin, Dihydrochloride, Cell Culture-Tested

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N7O5 · 2HCl
CAS Number:
Molecular Weight:
544.43
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

impurities

≤10 EU/mg Endotoxin (dry weight)

color

white to off-white

solubility

methanol: 20 mg/mL
water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C22H29N7O5.2ClH/c1-28(2)19-17-20(25-10-24-19)29(11-26-17)22-18(31)16(15(9-30)34-22)27-21(32)14(23)8-12-4-6-13(33-3)7-5-12;;/h4-7,10-11,14-16,18,22,30-31H,8-9,23H2,1-3H3,(H,27,32);2*1H/t14-,15+,16?,18+,22+;;/m0../s1

InChI key

MKSVFGKWZLUTTO-USYAMCSGSA-N

General description

An aminonucleoside antibiotic that acts as a prokaryotic and eukaryotic protein synthesis inhibitor. Resembles the aminoacyl-adenylyl terminus of aminoacyl-tRNA and competes for binding to the "A site" of the large ribosomal subunit. Its incorporation into a growing polypeptide results in termination of chain elongation and release of the nascent polypeptidyl-puromycin. Useful for selection of mammalian cell lines expressing a transfected pac gene (Streptomyces alboniger), whose product puromycin acetyltransferase, inactivates the drug via acetylation. Induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells. Recommended range for selection of transfected mammalian cells ranges from 1 to 30 µg/ml.
Protein synthesis inhibitor. Inhibits translation by causing premature release of nascent polypeptide chains. Induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells.
Puromycin, CAS 58-58-2, is a protein synthesis inhibitor that causes premature release of nascent polypeptide chains.

Biochem/physiol Actions

Primary Target
protein synthesis

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm pore-size filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Kaufman, S.H., et al. 1993. Cancer Res.53, 3976.
de la Luna, S. and Ortín, J. 1992. Methods Enzymol.216, 376.
Chow, S.C., et al. 1991. Exp. Cell Res. 216, 149.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baolong Xia et al.
Nature protocols, 15(10), 3478-3498 (2020-09-23)
Precise and efficient genome modifications provide powerful tools for biological studies. Previous CRISPR gene knockout methods in cell lines have relied on frameshifts caused by stochastic insertion/deletion in all alleles. However, this method is inefficient for genes with high copy
Jianxiang Lin et al.
Cell proliferation, 54(8), e13096-e13096 (2021-07-10)
PKM1 and PKM2, which are generated from the alternative splicing of PKM gene, play important roles in tumourigenesis and embryonic development as rate-limiting enzymes in glycolytic pathway. However, because of the lack of appropriate techniques, the specific functions of the
Marta Alenquer et al.
PLoS pathogens, 17(8), e1009772-e1009772 (2021-08-06)
Understanding SARS-CoV-2 evolution and host immunity is critical to control COVID-19 pandemics. At the core is an arms-race between SARS-CoV-2 antibody and angiotensin-converting enzyme 2 (ACE2) recognition, a function of the viral protein spike. Mutations in spike impacting antibody and/or
Juliana Gonçalves et al.
Cell reports. Medicine, 2(12), 100468-100468 (2021-12-08)
In view of the scarcity of data to guide decision making, we evaluated how BNT162b2 and mRNA-1273 vaccines affect the immune response in lactating women and the protective profile of breastmilk. Compared with controls, lactating women had a higher frequency
Jessica L Halle et al.
Frontiers in cell and developmental biology, 9, 690452-690452 (2021-08-17)
Preclinical models and in vitro experiments have provided valuable insight into the regulation of cancer-induced muscle wasting. Colon-26 (C26) tumor cells induce cachexia in mice, and conditioned media (CM) from these cells promotes myotube atrophy and catabolic signaling. While mechanical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service