Skip to Content
Merck
All Photos(1)

Key Documents

N1142

Sigma-Aldrich

Neomycin

solution, suitable for cell culture, BioReagent

Synonym(s):

Antibiotic 10676, Neomycin B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Product Name

Neomycin solution, with 10 mg/mL neomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture

biological source

Streptomyces fradiae

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

potency

8-12 mg per mL

packaging

pkg of 20ML

concentration

10 mg/mL in 0.9% NaCl (neomycin)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

clear

UniProt accession no.

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O)[C@H]2O[C@@H]3O[C@H](CO)[C@@H](O[C@H]4O[C@@H](CN)[C@@H](O)[C@H](O)[C@H]4N)[C@H]3O)[C@H](N)[C@@H](O)[C@@H]1O

InChI

1S/C23H46N6O13/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22/h5-23,30-36H,1-4,24-29H2/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+/m1/s1

InChI key

PGBHMTALBVVCIT-VCIWKGPPSA-N

Gene Information

human ... TERT(7015)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Neomycin solution has been used:
  • as a supplement of autoclaved acidified water for bone marrow transplantation[1]
  • for the selection of stable clones[2]
  • as a component of artificial sea water (ASWO) medium to remove bacteria from O. tauri cultures[3]

Biochem/physiol Actions

Neomycin is an aminoglycoside antibiotic, produced by Streptomyces containing a minimum of 85% neomycin B.[4] It acts as a selection agent for prokaryotic cells transformed using the neo selectable marker gene. Neomycin trisulfate is used to study the cytototoxic side effects of antibiotics, platelet-derived growth factor responses in certain fibroblasts and extraction of nuclear phosphatidylinositol 4,5-bisphosphate-interacting proteins. This product is recommended for use in cell culture applications at 5 mL/L.
Mode of action: This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.
Antimicrobial spectrum: Neomycin acts against both gram-positive and gram-negative bacteria

Caution

This solution is formulated to contain 10 mg/mL neomycin in 0.9% sodium chloride. It is stable at 37 °C for 5 days and should be otherwise stored at 2-8°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tudor Arvinte et al.
Journal of pharmaceutical and biomedical analysis, 175, 112742-112742 (2019-07-26)
The biosimilarity assessment of the physicochemical properties of high-concentration biopharmaceuticals is usually performed with measurements on diluted solutions, at concentrations below 1 mg/mL. In this study 13 orthogonal, spectroscopy and particle size determination methods were used to characterize the structure and
Long non-coding antisense RNA controls Uchl1 translation through an embedded SINEB2 repeat
Carrieri C, et al.
Nature, 491(7424), 454-454 (2012)
Metabolic profiling identifies trehalose as an abundant and diurnally fluctuating metabolite in the microalga Ostreococcus tauri
Hirth M, et al.
Metabolomics, 13(6), 68-68 (2017)
Tudor Arvinte et al.
Journal of pharmaceutical and biomedical analysis, 176, 112802-112802 (2019-08-26)
The physicochemical properties of Avastin® manufactured in the USA (Originator USA) and in Europe (Originator EU) and ABX-BEV, a bevacizumab biosimilar drug product candidate produced by Apobiologix Inc., were characterized at a clinically relevant concentration of 2 mg/mL following dilution of
Mandell, Douglas, and Bennett's Principles and Practice of Infectious Diseases (2014)

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service