Skip to Content
Merck
All Photos(1)

Key Documents

538907

Sigma-Aldrich

Prostaglandin F

Major uterine luteolytic prostaglandin.

Synonym(s):

Prostaglandin F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 7878

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

ethanol: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1C(C(C(C1)O)\C=C\C(O)CCCCC)C\C=C\CCCC(=O)O

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-19,21-23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4+,13-12+

InChI key

PXGPLTODNUVGFL-RTYMFESYSA-N

General description

Major uterine luteolytic prostaglandin.
Major uterine luteolytic prostaglandin. A pulmonary vasoconstrictor. Inhibits nitric oxide production by the uterine tissue and increases uterine contractility.

Biochem/physiol Actions

Primary Target
A pulmonary vasoconstrictor
Product does not compete with ATP.

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Dong, Y.L., et al. 1997. Am. J. Obstet. Gynecol. 177, 907.
Vincent, D.L., et al. 1986. Prostaglandins 31, 715.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service