Skip to Content
Merck
All Photos(1)

Key Documents

442579

Supelco

Endrin ketone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6O
CAS Number:
Molecular Weight:
380.91
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-30°C

SMILES string

Cl[C@@H]1[C@@]2(Cl)[C@H]3[C@@H]4C[C@@H]5[C@H]3[C@](Cl)(C2(Cl)Cl)[C@@]1(Cl)[C@@H]5C4=O

InChI

1S/C12H8Cl6O/c13-8-9(14)6-2-1-3(7(6)19)4-5(2)11(9,16)12(17,18)10(4,8)15/h2-6,8H,1H2/t2-,3+,4+,5-,6+,8+,9-,10+,11-/m1/s1

InChI key

IZHZFAQWVKBTSL-YTMKBKNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Endrin ketone is classified under the group of organochlorine pesticides (OCPs) used in agriculture for the control of pests and diseases.

Application

Endrin ketone may be used as an analytical reference standard for the determination of the analyte in environmental samples, water samples, vegetation samples and dairy samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a solid-phase microextraction gas chromatography with microelectron-capture detection method for a multiresidue analysis of pesticides in bovine milk.
Fernandez-Alvarez M, et al.
Analytica Chimica Acta, 617(1-2), 37-50 (2008)
Determination of organochlorine pesticides in ground water samples using solid-phase microextraction by gas chromatography-electron capture detection.
Junior JLR and Re-Poppi N
Talanta, 72(5), 1833-1841 (2007)
Application of ultrasound-assisted emulsification-micro-extraction for the analysis of organochlorine pesticides in waters.
Ozcan S, et al.
Water Research, 43(17), 4269-4277 (2009)
A sonication extraction method for the analysis of pyrethroid, organophosphate, and organochlorine pesticides from sediment by gas chromatography with electron-capture detection.
You J, et al.
Archives of Environmental Contamination and Toxicology, 47(2), 141-147 (2004)
Microwave-assisted extraction versus Soxhlet extraction in the analysis of 21 organochlorine pesticides in plants.
Barriada-Pereira M, et al.
Journal of Chromatography A, 1008(1), 115-122 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service