Skip to Content
Merck
All Photos(4)

Documents

P0424

Sigma-Aldrich

Prostaglandin F tris salt

≥99%, synthetic, powder

Synonym(s):

(5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt, PGF−Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5 · C4H11NO3
CAS Number:
Molecular Weight:
475.62
Beilstein:
4087514
EC Number:
MDL number:
UNSPSC Code:
12352401
eCl@ss:
42020658
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥99%

form

powder

color

white

solubility

H2O: 1 mg/mL
ethanol: 50 mg/mL

functional group

carboxylic acid
hydroxyl

shipped in

ambient

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.[H][C@]1(C\C=C/CCCC(O)=O)[C@@H](O)C[C@@H](O)[C@]1([H])\C=C\[C@@H](O)CCCCC

InChI

1S/C20H34O5.C4H11NO3/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25;5-4(1-6,2-7)3-8/h4,7,12-13,15-19,21-23H,2-3,5-6,8-11,14H2,1H3,(H,24,25);6-8H,1-3,5H2/b7-4-,13-12+;/t15-,16+,17+,18-,19+;/m0./s1

InChI key

IYGXEHDCSOYNKY-RZHHZEQLSA-N

Gene Information

human ... PTGFR(5737)

Looking for similar products? Visit Product Comparison Guide

General description

Aqueous solutions are stable for 30 days at 2-8°C and for several months frozen at −0°C in single use aliquots. Avoid repeated freeze/thaw cycles.

Biochem/physiol Actions

Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. A positive feedback loop between endometrial PG F and luteal oxytocin is responsible for completion of luteolysis.
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. Also stimulates contraction of smooth muscle, such as in bronchi, with possible involvement in asthma. This effect is at least partially mediated through tachykinin release.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Fuchs
Journal of steroid biochemistry, 27(4-6), 1073-1080 (1987-01-01)
The oxytocin-neurophysin gene is expressed in several nontraditional sites within the endocrine system. In the ovary its expression in the corpora lutea is initiated by ovulation. Ovarian oxytocin concentrations reach maximal levels around day 11 of luteal cycle and fall
J B Salom et al.
European journal of pharmacology, 203(1), 33-39 (1991-10-02)
We examined the effects of extracellular Ca2+ withdrawal and of Ca2+ entry blockers on goat cerebrovascular responses to prostaglandin F2 alpha (PGF2 alpha). We measured isometric tension in isolated middle cerebral arteries, and cerebral blood flow (CBF) in unanesthetized animals.
J W Barnard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 72(6), 2469-2474 (1992-06-01)
To characterize the interactions between prostaglandin F2 alpha and prostacyclin in controlling tone in the pulmonary circulation, isolated rat lungs were ventilated, perfused with blood, and subjected to challenge by prostaglandin F2 alpha in increasing doses. The pulmonary resistance was
M Szekeres et al.
Journal of vascular research, 35(5), 332-344 (1998-10-28)
The depedence of elastic and contractile properties on the caliber of small intramural coronary arteries was investigated in the rat in vitro. Different segments of the left anterior descending coronary artery branching system were prepared for microarteriography. The segments were
W Wuttke et al.
Steroids, 63(5-6), 299-305 (1998-06-10)
During the second half of the luteal phase, the human corpus luteum becomes responsive to regular luteinizing hormone (LH) pulses. These LH pulses stimulate progesterone secretion tonically, and during this tonic stimulation, additional LH-independent progesterone pulses occur, which are particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service