Skip to Content
Merck
All Photos(3)

Documents

A21207

Sigma-Aldrich

3-Acetylpyridine

98%

Synonym(s):

Methyl 3-pyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
107751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

220 °C (lit.)

mp

11-13 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccnc1

InChI

1S/C7H7NO/c1-6(9)7-3-2-4-8-5-7/h2-5H,1H3

InChI key

WEGYGNROSJDEIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matasha Dhar et al.
Nature communications, 7, 12938-12938 (2016-09-22)
Neurodegenerative lesions induce sprouting of new collaterals from surviving axons, but the extent to which this form of axonal remodelling alters brain functional structure remains unclear. To understand how collateral sprouting proceeds in the adult brain, we imaged post-lesion sprouting
Huawen Lin et al.
PLoS genetics, 6(9), e1001105-e1001105 (2010-09-15)
The essential coenzyme nicotinamide adenine dinucleotide (NAD+) plays important roles in metabolic reactions and cell regulation in all organisms. Bacteria, fungi, plants, and animals use different pathways to synthesize NAD+. Our molecular and genetic data demonstrate that in the unicellular
Iran Goudarzi et al.
European journal of pharmacology, 642(1-3), 56-65 (2010-07-20)
Electrophysiological dysfunction of Purkinje cells causes cerebellar ataxia. Recent studies indicated that 4-aminopyridine (4-AP) can prevent the attacks in patients with episodic ataxia type 2. However, the cellular mechanism(s) by which 4-AP might be beneficial for the improvement of motor
M Kaffashian et al.
Physiological research, 60(2), 355-365 (2010-12-01)
Alterations in the intrinsic properties of Purkinje cells (PCs) may contribute to the abnormal motor performance observed in ataxic rats. To investigate whether such changes in the intrinsic neuronal excitability could be attributed to the role of Ca(2+)-activated K(+) channels
Ai-Li Cai et al.
The European journal of neuroscience, 24(8), 2169-2176 (2006-10-18)
Zinc neurotoxicity has been demonstrated in ischemic, seizure, hypoglycemic, and trauma-induced neuronal death where Zn(2+) is thought to be synaptically released and taken up in neighbouring neurons, reaching toxic concentrations. We previously demonstrated that toxicity of extracellular Zn(2+) depended on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service