Skip to Content
Merck
All Photos(1)

Documents

354102

Sigma-Aldrich

Glutathione Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.84

usage

sufficient for 100 tests

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

do not freeze
protect from light

assay range

sensitivity: 5.0 μM
(RSH)

input

sample type cell lysate
sample type cell lysate
sample type tissue homogenate(s)
sample type tissue homogenate(s)

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

General description

A spectrophotometric assay kit that is specific for reduced glutathione (GSH) or total mercaptans. Absorbance is read at ~400 nm for GSH and ~356 nm for total mercaptens.
Note: 1 T = 1 test.
Oxidative stress occurs in most, if not all, human diseases. Our understanding of the role played by reactive oxygen species in the progression of pathological changes is still evolving. Convenient, accurate and reproducible methodologies to measure oxidative stress parameters have been lacking. Glutathione (γ-glutamylcysteinylglycine or GSH) is a naturally occuring tripeptide whose nucleophilic and reducing properties play a central role in metabolic pathways, as well as in the antioxidant system of most aerobic cells. GSH plays a critical role as a coenzyme with a variety of enzymes including, glutathione peroxidase, glutathione S-transferase, and thiol transferase. GSH also plays major roles in drug metabolism, calcium metabolism, the γ-glutamyl cycle, blood platelet, and membrane functions. In addition, GSH is crucial to a variety of life processes, including the detoxification of xenobiotics, maintenance of the -SH level of proteins, thiol-disulfide exchange, removal of hydroperoxides and free radicals, and amino acid transport across membranes. Physiological values of intracellular GSH generally range from 1 to 10 mM. Although many methods have been described for the assay of GSH, the reliable ones are labor intensive and not easy to use.

Components

A Chromogen, 30% NaOH, Buffer, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Specifications

Assay Time: 1.5 h

Principle

The Calbiochem® Glutathione Assay Kit makes it possible to assay for glutathione with only one sampling and one colorimetric measurement. A modification of this method makes it possible to assay other mercaptans. This alternate protocol is based on the measurement of substitution products that absorb maximally at 356 nm in the absence of reagent R2. Due to its ease of use, this method is well adapted for the quantification of glutathione and total mercaptans in a large number of biological samples. The main advantage of the method is that it does not require any enzyme as a reagent.

Preparation Note

Erythrocyte Lysate

1. Centrifuge a minimum of 500 l of whole blood at 2500 X g at 4°C for 5 min.
2. Discard plasma supernatant. If not assayed immediately, store erythrocyte pellet at -70°C (erythrocyte pellet can be stored at -70°C for 15 days).
3. Resuspend erythrocyte pellet in 4 volumes of MPA working solution, 0-4°C.
4. Thoroughly mix and centrifuge at 3000g at 4°C for 10 min.
5. Collect the upper clear aqueous layer and keep at 0-4°C for the assay (within 1 h).

Liver Homogenates

1. Wash tissue in 0.9% NaCl solution.
2. Blot tissue on paper and weigh.
3. Mince tissue in ice-cold MPA working solution.
4. Homogenize minced tissue.
5. Centrifuge homogenate at 3000 x g, 4°C for 10 min.
6. Collect the upper clear aqueous layer* and keep at 0-4°C for the assay (within 1 h).
*Cloudy supernatant should be filtered through 0.2 m filters.

Hepatocyte Lysates

1. Resuspend hepatocyte* pellet, from rats or mice, in 500 l of ice-cold MPA working solution.
2. Homogenize cell suspension.
3. Centrifuge homogenate at 3000 x g, 4°C for 10 min.
4. Collect the upper clear aqeous layer and keep at 0-4°C for the assay (within 1 h).
*Approximately 2.5-3.5 x 106 cells are used (5-8 mg of total protein).

Storage and Stability

Upon arrival store the entire kit contents at 4°C.

All solutions of reagents and buffer have been tightly sealed and are stable if properly stored between 4°C. Do Not Freeze. Protect from light.

After removing the required amount of each reagent for immediate use, all bottles should be tightly closed and stored between 2-4°C. Do not leave the reagent bottles open or at room temperature.

Analysis Note

Measurement of Total Mercaptans (RSH): Assay at 356 nm

The Calbiochem Glutathione Assay Kit can be used for the measurement of other mercaptans (RSH), which include GSH. The assay is carried out in the absence of reagent R2 at 356 nm. The absorbance at 356 nm is a linear function of [RSH] concentration in the sample, but it is not GSH-specific. If the sample essentially contains GSH and a single other mercaptan i.e., N-acetylcysteine, these two mercaptans can be assayed by using the same single sample for two measurements. The first measurement is made at 356 nm before the addition of reagent R2 as described below, and the second measurement is made, after the addition of R2, at 400 nm.

Adjust the spectrophotometer absorbance to zero at 356 nm with buffer only (solution 3). The reaction mixture is prepared as in Section 3, with the omission of step 4. The absorbance (A) is measured at 356 nm.

A standard curve must be prepared with the corresponding mercaptan in Table 2, to calculate the concentration. Figure 2 gives two examples of standard curves obtained with glutathione and Nacetylcysteine at 356 nm.

GSH Concentration
The calculation is based on the following equation:
[GSH] = {(A-A0)/(E x l)} x D
where:
[GSH] is the initial glutathione concentration in the sample, expressed as molar concentration.

A and A0 are the absorbances measured in the presence and in the absence of sample, respectively.

E is the apparent molar extinction coefficient of the product measured at 400 nm.

l is the optical path (cm).

D is the dilution factor of the sample.

Total Mercaptan Concentration
The calculation is based on the modified version of the equation used for GSH:
[total RSH] = { (A - A0) / (E x I) } x D
where :
[total RSH] is the concentration of total mercaptan in the sample.

A and A0 are the absorbances measured in the presence and in the absence of sample, respectively.

E is the apparent molar extinction coefficient of the product measured at 356 nm.
I is the optical path (cm).

D is the dilution factor of the sample.

Note:
  • Do not add reagents R1 and R2 in reverse order.
  • The temperature, (25 ± 3°C), should be kept constant throughout the experiment.
  • The final reaction volume (1 ml) should not vary from one measurement to another.
  • The absorbance at 400 nm is proportional to glutathione concentration. It is stable for min, provided that the reaction mixture is kept in the dark.
  • The sensitivity of the assay for mercaptans at 356 nm is of the same order of magnitude as that of glutathione at 400 nm as shown in Table 2.
Sensitivity: From 30 repeated measurements performed on the control ([RSH] = 0) on the same day, under the same experimental conditions, the detection limit of the assay was 5 µmol/l in the final reaction mixture (spectrophotometric cuvette). For example, using the maximum volume of 300 µl of a sample would therefore give a detection limit for glutathione of about 17 µmol/l.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Ji, L.L., and Fu, R. 1992. J. Appl. Physiol.72, 549.
Anderson M.E. 1989. Enzymatic and chemical methods for the determination of
Glutathione; In: Glutathione: chemical, biochemical and medical aspects, Vol. A, Dolphin D., Poulson R. and Avramovic O. Eds., John Wiley and Sons, pp. 339-365.
Dolphin D., et al. 1989. Glutathione: Chemical, Biochemical and Medical Aspects, Vols A & B, J. Wiley and Sons.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgian T Mustata et al.
Diabetes, 54(2), 517-526 (2005-01-29)
We tested the hypothesis that green tea prevents diabetes-related tissue dysfunctions attributable to oxidation. Diabetic rats were treated daily with tap water, vitamins C and E, or fresh Japanese green tea extract. After 12 months, body weights were decreased, whereas
Suh-Ching Yang et al.
The British journal of nutrition, 92(2), 209-215 (2004-08-31)
The purpose of the present study was to evaluate the effects of beta-carotene on the cell viability and antioxidant status of hepatocytes from chronically ethanol-fed rats. Rats in the ethanol group were given an ethanol-containing liquid diet that provided 36
Wang-teng Lin et al.
Acta pharmacologica Sinica, 26(8), 992-999 (2005-07-26)
To assess the effects of L-arginine (L-Arg) supplementation on pulmonary oxidative stress and antioxidant defenses in rats after exhaustive exercise. Rats were randomly divided into four groups: sedentary control (SC), sedentary control with L-Arg treatment (SC+Arg), exhaustive exercise with control
William Y Boadi et al.
Drug and chemical toxicology, 39(3), 239-247 (2016-04-12)
Many studies have shown that cellular redox potential is largely determined by glutathione (GSH), which accounts for more than 90% of cellular nonprotein thiols. The aim of this study was to delineate the effect of three flavonoids - namely, quercetin
Glutathione Biosynthesis Influences Replicative Longevity in Saccharomyces Cerevisiae.
Sylvie M Van Zandycke et al.
TheScientificWorldJournal, 1, 133-133 (2001-10-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service