Skip to Content
Merck
All Photos(1)

Documents

AP130P

Sigma-Aldrich

Goat Anti-Mouse IgG & IgM Antibody, HRP conjugate

0.8 mg/mL, Chemicon®

Synonym(s):

Goat Anti-Mouse IgG Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

0.8 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is the most abundant immunoglobulin in the human serum. IgM constitutes 5-10% of total serum immunoglobulins. These immunoglobulins comprise four polypeptide chains with two heavy (H) chains and two light (L) chains. These polypeptide chains are inter-linked by disulfide bonds. IgM is a highly conserved antibody in vertebrates. IgM exists as a pentamer. Both IgG and IgM are secreted by B cells.

Application

Goat Anti-Mouse IgG & IgM Antibody, HRP conjugate has been used in western blotting(1:400) and immunofluorescence (1:500)

Biochem/physiol Actions

IgG facilitates antibody-dependent cell-mediated cytotoxicity (ADCC). It also aids in the activation of the classical pathway of the complement system. IgG neutralizes virus particles and toxins. IgM, an antibody that is expressed early during an immune response, aids in engulfing apoptotic cells. It exhibits polyreactive and autoreactive functionality. IgM induces tissue homeostasis by mediating the clearance of tissue-based molecules. Lower levels of IgM in serum are associated with increased autoimmune response and a higher risk of developing infections.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bojana Mirković et al.
Oncotarget, 6(22), 19027-19042 (2015-04-08)
Cathepsin B is a ubiquitously expressed lysosomal cysteine protease that participates in protein turnover within lysosomes. However, its protein and activity levels have been shown to be increased in cancer. Cathepsin B endopeptidase activity is involved in the degradation of
Chapter 12 - Immunoglobulin Titers and Immunoglobulin Subtypes
Gleason CA and Devaskar SU
Acanthamoeba: Biology and Pathogenesis (2011)
Dusan Bencina et al.
Avian pathology : journal of the W.V.P.A, 34(6), 463-472 (2006-03-16)
Maternal antibodies can protect avian embryos against vertically transmitted pathogens during embryogenesis and also young birds after hatching. In contrast to the well-known transfer of maternal immunoglobulin (Ig) G (also termed IgY) from the yolk to embryonic blood, information about
Mari H Savolainen et al.
Neurobiology of disease, 68, 1-15 (2014-04-22)
The misfolding and aggregation of α-synuclein (aSyn) eventually lead to an accumulation of toxic forms that disturb normal neuronal function and result in cell death. aSyn rich inclusions are seen in Parkinson's disease, dementia with Lewy bodies and other synucleinopathies.
Candida J Rebello et al.
Obesity (Silver Spring, Md.), 27(1), 103-111 (2018-12-07)
Naringenin, a citrus flavonoid, prevents diet-induced weight gain and improves glucose and lipid metabolism in rodents. There is evidence that naringenin activates brown fat and increases energy expenditure in mice, but little is known about its effects in humans. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service