Skip to Content
Merck
All Photos(2)

Key Documents

1.08331

Supelco

Toluene

for spectroscopy Uvasol®

Synonym(s):

Toluol, Methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
NACRES:
NA.03
Assay:
≥99.9% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

3.2 (vs air)

Quality Level

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0001 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

285 nm, ≥15%
290 nm, ≥60%
300 nm, ≥80%
310 nm, ≥90%
335 nm, ≥96%
350 nm, ≥98%

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

density

0.865 g/mL at 25 °C (lit.)

UV absorption

λ: 285 nm Amax: ≤0.82
λ: 290 nm Amax: ≤0.22
λ: 300 nm Amax: ≤0.10
λ: 310 nm Amax: ≤0.05
λ: 335 nm Amax: ≤0.02
λ: 350 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service